Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPL4 SIRNA Assays ›
          Gene Symbol:
          Rpl4
          Gene Name:
          ribosomal protein L4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 69121682 - 69126805 on Build Rnor_6.0
          UniGene ID:
          Rn.1133
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 192113

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rpl4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022510.1 NP_071955.1 5 557
          GenBank mRNA BC063811.1 - 5 573 6919340
          BC081801.1 - 5 528 7192390
          FQ209448.1 - 4 411
          FQ209532.1 - 4 411
          FQ209571.1 - 4 411
          FQ212557.1 - 4 373
          FQ212566.1 - 4 411
          FQ212656.1 - 4 411
          FQ212896.1 - 4 411
          FQ212979.1 - 2 203
          FQ213045.1 - 4 411
          FQ213454.1 - 5 562
          FQ215017.1 - 4 367
          FQ215042.1 - 4 411
          FQ215264.1 - 4 411
          FQ215283.1 - 4 411
          FQ215533.1 - 4 411
          FQ215586.1 - 4 411
          FQ215649.1 - 4 411
          FQ216342.1 - 4 411
          FQ216573.1 - 4 411
          FQ216754.1 - 4 411
          FQ216792.1 - 4 411
          FQ218195.1 - 4 411
          FQ219327.1 - 4 411
          FQ219561.1 - 4 411
          FQ219701.1 - 4 411
          FQ219716.1 - 4 411
          FQ220231.1 - 4 409
          FQ220328.1 - 2 233
          FQ221130.1 - 4 373
          FQ226216.1 - 4 411
          FQ228709.1 - 4 411
          FQ228734.1 - 5 562
          FQ228797.1 - 4 411
          FQ228847.1 - 4 411
          FQ228910.1 - 4 411
          FQ228929.1 - 4 411
          FQ228930.1 - 4 411
          FQ229086.1 - 4 411
          FQ229087.1 - 4 411
          FQ229116.1 - 4 411
          FQ229723.1 - 2 151
          FQ229849.1 - 4 370
          FQ231653.1 - 4 411
          FQ232105.1 - 4 411
          FQ232562.1 - 4 411
          FQ232771.1 - 4 411
          FQ233898.1 - 4 411
          FQ234293.1 - 4 411
          FQ234521.1 - 4 411
          FQ234619.1 - 4 411
          X82180.1 - 5 557

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: AK088054, AK088279.

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0