Search
Search

Gng5| Gene Symbol: Gng5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024377.1 | NP_077353.1 | 3 | 279 | ||
| GenBank mRNA | BC058469.1 | - | 3 | 363 | 6918279 | |
| DQ120493.1 | - | 3 | 224 | |||
| DQ120494.1 | - | 3 | 224 | |||
| M95780.1 | - | 3 | 279 | |||