Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CRH SIRNA Assays ›
Gene Symbol:
Crh
Gene Name:
corticotropin releasing hormone
Gene Aliases:
CRF
Chromosome Location:
Chr.2: 104459999 - 104461863 on Build Rnor_6.0
UniGene ID:
Rn.10349
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
194924
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Crh
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031019.1
NP_112281.1
2
706
GenBank mRNA
X03036.1
-
2
704
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_000756, BC002599, BC011031, AK075431, BT007453.
This siRNA also targets the following transcripts in Mouse: NM_205769, BC119036, BC116921.
Panther Classification:
Molecular Function -
peptide hormone
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
synaptic transmission, dopaminergic
glucocorticoid biosynthetic process
inflammatory response
female pregnancy
learning or memory
steroid metabolic process
positive regulation of cell proliferation
associative learning
hormone-mediated apoptotic signaling pathway
positive regulation of gene expression
negative regulation of gene expression
negative regulation of norepinephrine secretion
positive regulation of cell death
regulation of serotonin secretion
diterpenoid metabolic process
hypothalamus development
lung development
adrenal gland development
positive regulation of cAMP biosynthetic process
negative regulation of epinephrine secretion
negative regulation of luteinizing hormone secretion
locomotory exploration behavior
positive regulation of insulin secretion involved in cellular response to glucose stimulus
response to immobilization stress
response to cocaine
response to drug
response to estrogen
response to ethanol
response to ether
negative regulation of blood pressure
response to pain
ion homeostasis
response to corticosterone
positive regulation of corticotropin secretion
positive regulation of cortisol secretion
long-term synaptic potentiation
positive regulation of digestive system process
negative regulation of cell death
negative regulation of glucagon secretion
cellular response to cocaine
cellular response to dexamethasone stimulus
positive regulation of calcium ion import
regulation of N-methyl-D-aspartate selective glutamate receptor activity
positive regulation of corticosterone secretion
positive regulation of behavioral fear response
hormone activity
protein binding
corticotropin-releasing hormone receptor 1 binding
corticotropin-releasing hormone receptor 2 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-48z6k:80/100.66.78.162:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0