Search
Search

CASC3| Gene Symbol: CASC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007359.4 | NP_031385.2 | 14 | 3901 | ||
| XM_005257163.1 | XP_005257220.1 | 14 | 3674 | |||
| GenBank mRNA | AK096126.1 | - | 3 | 2526 | ||
| BC044656.1 | - | 14 | 3784 | 5547561 | ||
| BC050526.1 | - | 14 | 3804 | 5561860 | ||
| X80199.2 | - | 14 | 3839 | |||