Search
Search

Becn1| Gene Symbol: Becn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034117.1 | NP_001029289.1 | 9 | 1055 | ||
| NM_053739.2 | NP_446191.1 | 9 | 1077 | |||
| GenBank mRNA | AY033824.1 | - | 8 | 853 | ||
| BC074011.1 | - | 9 | 942 | 7100185 | ||