Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FCGR2A SIRNA Assays ›
          Gene Symbol:
          Fcgr2a LOC100911825 LOC103693683 LOC103694908 LOC498276
          Gene Name:
          Fc fragment of IgG, low affinity IIa, receptor
          low affinity immunoglobulin gamma Fc region receptor III-like
          low affinity immunoglobulin gamma Fc region receptor III
          low affinity immunoglobulin gamma Fc region receptor III-like
          Fc gamma receptor II beta
          Gene Aliases:
          Fcgr3, Fcgr3a
          -
          -
          -
          -
          Chromosome Location:
          Chr.13: 91146879 - 91163694 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 91219838 - 91237277 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 89420835 - 89440644 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906965: 601 - 13578 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 91168972 - 91181954 on Build Rnor_6.0
          UniGene ID:
          Rn.154415
          -
          Rn.6050
          -
          Rn.6050
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID 195783

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fcgr2a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053843.2 NP_446295.2 6 1039
          XM_006250303.2 XP_006250365.1 6 1092
          XM_006250305.2 XP_006250367.1 6 1191
          GenBank mRNA BC060534.1 - 816 6886727
          L08446.1 - 6 1039
          M32062.1 - 852
          M64368.1 - 832
          M64369.1 - 832
          M64370.1 - 832
          Gene Symbol: LOC100911825
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006250308.2 XP_006250370.1 5 860
          XM_008769774.1 XP_008767996.1 5 860
          XM_008769775.1 XP_008767997.1 6 1085
          XM_008769776.1 XP_008767998.1 6 893
          XM_008769777.1 XP_008767999.1 4 839
          Gene Symbol: LOC103693683
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008769767.1 XP_008767989.1 6 881
          XM_008769768.1 XP_008767990.1 6 1086
          XM_008769769.1 XP_008767991.1 6 1673
          XM_008769770.1 XP_008767992.1 6 1655
          XM_008769771.1 XP_008767993.1 5 862
          XM_008769772.1 XP_008767994.1 6 1266
          XM_008769773.1 XP_008767995.1 5 844
          Gene Symbol: LOC103694908
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008773904.1 XP_008772126.1 860
          XM_008773905.1 XP_008772127.1 861
          XM_008773906.1 XP_008772128.1 988
          Gene Symbol: LOC498276
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135992.1 NP_001129464.1 899
          XM_008769748.1 XP_008767970.1 860
          XM_008769749.1 XP_008767971.1 860
          XM_008769750.1 XP_008767972.1 984
          GenBank mRNA BC159405.1 - 899 7459664

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          nerve development
          Fc-gamma receptor signaling pathway
          IgG receptor activity
          IgG binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline