Search
Search

B3gat1| Gene Symbol: B3gat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054003.1 | NP_446455.1 | 5 | 1047 | ||
| XM_006242733.2 | XP_006242795.1 | 5 | 1319 | |||
| XM_006242734.2 | XP_006242796.1 | 5 | 1308 | |||
| GenBank mRNA | D88035.1 | - | 5 | 1047 | ||