Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GALK2 SIRNA Assays ›
          Gene Symbol:
          GALK2
          Gene Name:
          galactokinase 2
          Gene Aliases:
          GK2
          Chromosome Location:
          Chr.15: 49155757 - 49367883 on Build GRCh38
          UniGene ID:
          Hs.122006
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 196

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 417.00
          Your Price
          Online offer:
          417.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GALK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001556.2 NP_001001556.1 2 282
          NM_001289030.1 NP_001275959.1 3 514
          NM_001289031.1 NP_001275960.1 2 367
          NM_002044.3 NP_002035.1 2 384
          XM_005254279.3 XP_005254336.1 2 173
          XM_005254280.3 XP_005254337.1 2 264
          XM_005254284.3 XP_005254341.1 2 264
          XM_006720461.3 XP_006720524.1 2 173
          XM_006720462.3 XP_006720525.1 2 154
          XM_006720463.3 XP_006720526.1 2 153
          XM_006720464.3 XP_006720527.1 2 153
          XM_011521441.2 XP_011519743.1 2 232
          XM_017022062.1 XP_016877551.1 2 154
          XM_017022063.1 XP_016877552.1 2 264
          XM_017022065.1 XP_016877554.1 2 284
          XM_017022066.1 XP_016877555.1 2 305
          GenBank mRNA AF461816.1 - 2 262
          AK098049.1 - 2 123
          AK301361.1 - 2 137
          AK316440.1 - 3 248
          AL832745.1 - 2 261
          BC005141.2 - 2 152 3504930
          BI766538.1 - 2 174
          BQ921903.1 - 2 95
          BT006901.1 - 2 77
          DB457948.1 - 2 384
          DC298656.1 - 2 374
          DC390737.1 - 3 483
          M84443.1 - 2 97

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          carbohydrate kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          galactose metabolic process
          carbohydrate phosphorylation
          galactokinase activity
          ATP binding
          N-acetylgalactosamine kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline