Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL10 SIRNA Assays ›
Gene Symbol:
Il10
Gene Name:
interleukin 10
Gene Aliases:
IL10X
Chromosome Location:
Chr.13: 47739187 - 47743392 on Build Rnor_6.0
UniGene ID:
Rn.9868
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
197131
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Il10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012854.2
NP_036986.2
5
517
GenBank mRNA
L02926.1
-
5
517
X60675.1
-
560
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cytokine production
response to molecule of bacterial origin
negative regulation of cytokine secretion involved in immune response
negative regulation of chronic inflammatory response to antigenic stimulus
positive regulation of B cell apoptotic process
inflammatory response
immune response
aging
response to organic substance
regulation of gene expression
response to activity
response to inactivity
negative regulation of myeloid dendritic cell activation
negative regulation of B cell proliferation
response to lipopolysaccharide
negative regulation of interferon-gamma production
negative regulation of interleukin-12 production
negative regulation of interleukin-6 production
negative regulation of tumor necrosis factor production
receptor biosynthetic process
response to insulin
response to carbon monoxide
response to drug
negative regulation of tumor necrosis factor biosynthetic process
defense response to bacterium
negative regulation of apoptotic process
negative regulation of growth of symbiont in host
negative regulation of nitric oxide biosynthetic process
positive regulation of MHC class II biosynthetic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JAK-STAT cascade
positive regulation of cytokine secretion
negative regulation of inflammatory response
negative regulation of membrane protein ectodomain proteolysis
positive regulation of sequence-specific DNA binding transcription factor activity
response to glucocorticoid
regulation of sensory perception of pain
branching involved in labyrinthine layer morphogenesis
cellular response to lipopolysaccharide
cellular response to estradiol stimulus
regulation of complement-dependent cytotoxicity
negative regulation of endothelial cell apoptotic process
cytokine activity
interleukin-10 receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qb7lb:80/100.66.77.199:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0