Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AHSG SIRNA Assays ›
Gene Symbol:
Ahsg
Gene Name:
alpha-2-HS-glycoprotein
Gene Aliases:
Aa2-066, pp63
Chromosome Location:
Chr.11: 81711269 - 81717594 on Build Rnor_6.0
UniGene ID:
Rn.32083
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
197249
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ahsg
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012898.4
NP_037030.1
1
183
GenBank mRNA
AY325170.1
-
819
BC091118.1
-
1
161
7368203
FQ209390.1
-
1
180
FQ209416.1
-
1
182
FQ209417.1
-
1
182
FQ209422.1
-
1
182
FQ209445.1
-
1
182
FQ209451.1
-
1
182
FQ209502.1
-
1
182
FQ209536.1
-
1
182
FQ209545.1
-
1
182
FQ209557.1
-
1
182
FQ209568.1
-
1
182
FQ209581.1
-
1
182
FQ209585.1
-
1
182
FQ209588.1
-
1
182
FQ209604.1
-
1
182
FQ209612.1
-
1
182
FQ209617.1
-
1
182
FQ209654.1
-
1
182
FQ209655.1
-
1
180
FQ209665.1
-
1
182
FQ209680.1
-
1
182
FQ209720.1
-
1
182
FQ209759.1
-
1
182
FQ209767.1
-
1
191
FQ209793.1
-
1
182
FQ209828.1
-
1
137
FQ209858.1
-
1
180
FQ209920.1
-
1
182
FQ209985.1
-
1
182
FQ210030.1
-
1
177
FQ210033.1
-
1
182
FQ210056.1
-
1
180
FQ210082.1
-
1
182
FQ210085.1
-
1
180
FQ210120.1
-
1
181
FQ210139.1
-
1
182
FQ210141.1
-
1
182
FQ210159.1
-
1
182
FQ210164.1
-
1
180
FQ210170.1
-
1
182
FQ210199.1
-
1
182
FQ210228.1
-
1
182
FQ210239.1
-
1
182
FQ210242.1
-
1
179
FQ210249.1
-
1
182
FQ210260.1
-
1
182
FQ210304.1
-
1
182
FQ210382.1
-
1
182
FQ210396.1
-
1
182
FQ210402.1
-
1
182
FQ210405.1
-
1
182
FQ210452.1
-
1
183
FQ210457.1
-
1
182
FQ210498.1
-
1
182
FQ210504.1
-
1
182
FQ210525.1
-
1
181
FQ210562.1
-
1
182
FQ210566.1
-
1
182
FQ210623.1
-
1
182
FQ210700.1
-
1
182
FQ210709.1
-
1
181
FQ210731.1
-
1
180
FQ210738.1
-
1
182
FQ210739.1
-
1
182
FQ210760.1
-
1
180
FQ210764.1
-
1
182
FQ210871.1
-
1
182
FQ210891.1
-
1
182
FQ210896.1
-
1
180
FQ210953.1
-
1
182
FQ210957.1
-
1
188
FQ210983.1
-
1
182
FQ211020.1
-
1
182
FQ211045.1
-
1
179
FQ211090.1
-
1
182
FQ211092.1
-
1
182
FQ211115.1
-
1
182
FQ211177.1
-
1
182
FQ211179.1
-
1
182
FQ211283.1
-
1
182
FQ211299.1
-
1
182
FQ211301.1
-
1
182
FQ211303.1
-
1
182
FQ211304.1
-
1
182
FQ211368.1
-
1
182
FQ211379.1
-
1
182
FQ211843.1
-
1
182
FQ217075.1
-
1
180
FQ218089.1
-
1
182
FQ218223.1
-
1
181
FQ218262.1
-
1
182
FQ218269.1
-
1
182
FQ218332.1
-
1
182
FQ218341.1
-
1
182
FQ218342.1
-
1
180
FQ218355.1
-
1
181
FQ218373.1
-
1
182
FQ218390.1
-
1
182
FQ218393.1
-
1
182
FQ218400.1
-
1
182
FQ218424.1
-
1
180
FQ218436.1
-
1
180
FQ218463.1
-
1
180
FQ218464.1
-
1
182
FQ218469.1
-
1
180
FQ218479.1
-
1
182
FQ218480.1
-
1
182
FQ218487.1
-
1
182
FQ218489.1
-
1
182
FQ218500.1
-
1
182
FQ218502.1
-
1
182
FQ218509.1
-
1
180
FQ218531.1
-
1
182
FQ218563.1
-
1
180
FQ218569.1
-
1
180
FQ218570.1
-
1
182
FQ218580.1
-
1
182
FQ218585.1
-
1
182
FQ218608.1
-
1
182
FQ218609.1
-
1
182
FQ218630.1
-
1
178
FQ218648.1
-
1
180
FQ218657.1
-
1
182
FQ218695.1
-
1
182
FQ218713.1
-
1
180
FQ218744.1
-
1
164
FQ218765.1
-
1
182
FQ218787.1
-
1
184
FQ218796.1
-
1
182
FQ218800.1
-
1
182
FQ218813.1
-
1
269
FQ218846.1
-
1
182
FQ218858.1
-
1
179
FQ218862.1
-
1
182
FQ218872.1
-
1
182
FQ218877.1
-
1
180
FQ218889.1
-
1
182
FQ218890.1
-
1
182
FQ218892.1
-
1
182
FQ218907.1
-
1
182
FQ218912.1
-
1
182
FQ218931.1
-
1
181
FQ218948.1
-
1
182
FQ218951.1
-
1
182
FQ218970.1
-
1
180
FQ218974.1
-
1
183
FQ218975.1
-
1
182
FQ218980.1
-
1
182
FQ218981.1
-
1
180
FQ219018.1
-
1
182
FQ219028.1
-
1
182
FQ219033.1
-
1
182
FQ219042.1
-
1
181
FQ219060.1
-
1
182
FQ219061.1
-
1
182
FQ219086.1
-
1
182
FQ219094.1
-
1
182
FQ219099.1
-
1
182
FQ219103.1
-
1
182
FQ219110.1
-
1
182
FQ219121.1
-
1
182
FQ219129.1
-
1
180
FQ219130.1
-
1
178
FQ219141.1
-
1
182
FQ219164.1
-
1
182
FQ219173.1
-
1
182
FQ219194.1
-
1
182
FQ219203.1
-
1
182
FQ219221.1
-
1
182
FQ219232.1
-
1
188
FQ219235.1
-
1
182
FQ219239.1
-
1
182
FQ219251.1
-
1
182
FQ219274.1
-
1
182
FQ219295.1
-
1
182
FQ219300.1
-
1
182
FQ219311.1
-
1
182
FQ219320.1
-
1
182
FQ219326.1
-
1
182
FQ219358.1
-
1
181
FQ219363.1
-
1
184
FQ219364.1
-
1
182
FQ219369.1
-
1
180
FQ219370.1
-
1
182
FQ219377.1
-
1
182
FQ219388.1
-
1
182
FQ219400.1
-
1
182
FQ219406.1
-
1
182
FQ219412.1
-
1
182
FQ219419.1
-
1
180
FQ219486.1
-
1
177
FQ219487.1
-
1
182
FQ219488.1
-
1
182
FQ219498.1
-
1
180
FQ219536.1
-
1
182
FQ219546.1
-
1
182
FQ219551.1
-
1
182
FQ219552.1
-
1
180
FQ219558.1
-
1
182
FQ219565.1
-
1
182
FQ219587.1
-
1
180
FQ219606.1
-
1
182
FQ219607.1
-
1
180
FQ219608.1
-
1
180
FQ219617.1
-
1
182
FQ219643.1
-
1
182
FQ219653.1
-
1
181
FQ219660.1
-
1
182
FQ219663.1
-
1
182
FQ219668.1
-
1
183
FQ219672.1
-
1
182
FQ219688.1
-
1
180
FQ219691.1
-
1
182
FQ219726.1
-
1
182
FQ219755.1
-
1
182
FQ219769.1
-
1
182
FQ219770.1
-
1
182
FQ219772.1
-
1
181
FQ219773.1
-
1
182
FQ219774.1
-
1
182
FQ222367.1
-
1
180
M29758.1
-
1
183
X63446.1
-
1
141
Back To Top
More Information
Panther Classification:
Molecular Function -
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
ossification
protein complex assembly
acute-phase response
male gonad development
negative regulation of endopeptidase activity
cerebral cortex development
negative regulation of cell growth
negative regulation of bone mineralization
organ regeneration
cellular response to insulin stimulus
negative regulation of phosphorylation
positive regulation of bone resorption
negative regulation of insulin receptor signaling pathway
regulation of inflammatory response
positive regulation of phagocytosis
negative regulation of protein tyrosine kinase activity
cysteine-type endopeptidase inhibitor activity
kinase inhibitor activity
receptor signaling protein tyrosine kinase inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline