Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AHSG SIRNA Assays ›
          Gene Symbol:
          Ahsg
          Gene Name:
          alpha-2-HS-glycoprotein
          Gene Aliases:
          Aa2-066, pp63
          Chromosome Location:
          Chr.11: 81711269 - 81717594 on Build Rnor_6.0
          UniGene ID:
          Rn.32083
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 197250

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ahsg
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012898.4 NP_037030.1 1 251
          GenBank mRNA AY325170.1 - 887
          BC091118.1 - 1 229 7368203
          FQ209390.1 - 1 248
          FQ209416.1 - 1 250
          FQ209417.1 - 1 250
          FQ209422.1 - 1 250
          FQ209445.1 - 1 250
          FQ209451.1 - 1 250
          FQ209502.1 - 1 250
          FQ209536.1 - 1 250
          FQ209545.1 - 1 250
          FQ209557.1 - 1 250
          FQ209568.1 - 1 250
          FQ209581.1 - 1 250
          FQ209585.1 - 1 250
          FQ209588.1 - 1 250
          FQ209604.1 - 1 250
          FQ209612.1 - 1 250
          FQ209617.1 - 1 250
          FQ209654.1 - 1 250
          FQ209655.1 - 1 248
          FQ209665.1 - 1 250
          FQ209680.1 - 1 250
          FQ209720.1 - 1 250
          FQ209767.1 - 1 259
          FQ209793.1 - 1 250
          FQ209858.1 - 1 248
          FQ209920.1 - 1 250
          FQ209985.1 - 1 250
          FQ210030.1 - 1 245
          FQ210033.1 - 1 250
          FQ210056.1 - 1 248
          FQ210082.1 - 1 250
          FQ210085.1 - 1 248
          FQ210120.1 - 1 249
          FQ210139.1 - 1 250
          FQ210141.1 - 1 250
          FQ210159.1 - 1 250
          FQ210164.1 - 1 248
          FQ210170.1 - 1 250
          FQ210199.1 - 1 250
          FQ210228.1 - 1 250
          FQ210239.1 - 1 250
          FQ210242.1 - 1 247
          FQ210249.1 - 1 250
          FQ210260.1 - 1 250
          FQ210304.1 - 1 250
          FQ210382.1 - 1 250
          FQ210396.1 - 1 250
          FQ210402.1 - 1 250
          FQ210405.1 - 1 250
          FQ210452.1 - 1 251
          FQ210457.1 - 1 250
          FQ210498.1 - 1 250
          FQ210504.1 - 1 250
          FQ210525.1 - 1 249
          FQ210562.1 - 1 250
          FQ210566.1 - 1 250
          FQ210623.1 - 1 250
          FQ210700.1 - 1 250
          FQ210709.1 - 1 249
          FQ210731.1 - 1 248
          FQ210738.1 - 1 250
          FQ210760.1 - 1 248
          FQ210764.1 - 1 250
          FQ210871.1 - 1 250
          FQ210891.1 - 1 250
          FQ210896.1 - 1 248
          FQ210953.1 - 1 250
          FQ210957.1 - 1 256
          FQ210983.1 - 1 250
          FQ211020.1 - 1 250
          FQ211045.1 - 1 247
          FQ211090.1 - 1 250
          FQ211092.1 - 1 250
          FQ211115.1 - 1 250
          FQ211177.1 - 1 250
          FQ211179.1 - 1 250
          FQ211283.1 - 1 250
          FQ211299.1 - 1 250
          FQ211301.1 - 1 250
          FQ211303.1 - 1 250
          FQ211304.1 - 1 250
          FQ211368.1 - 1 250
          FQ211379.1 - 1 250
          FQ211843.1 - 1 250
          FQ217075.1 - 1 248
          FQ218089.1 - 1 250
          FQ218223.1 - 1 249
          FQ218236.1 - 1 250
          FQ218262.1 - 1 250
          FQ218264.1 - 1 250
          FQ218269.1 - 1 250
          FQ218332.1 - 1 250
          FQ218341.1 - 1 250
          FQ218342.1 - 1 248
          FQ218355.1 - 1 249
          FQ218373.1 - 1 250
          FQ218390.1 - 1 250
          FQ218393.1 - 1 250
          FQ218400.1 - 1 250
          FQ218424.1 - 1 248
          FQ218436.1 - 1 248
          FQ218463.1 - 1 248
          FQ218464.1 - 1 250
          FQ218469.1 - 1 248
          FQ218479.1 - 1 250
          FQ218480.1 - 1 250
          FQ218487.1 - 1 250
          FQ218489.1 - 1 250
          FQ218493.1 - 1 250
          FQ218500.1 - 1 250
          FQ218502.1 - 1 250
          FQ218509.1 - 1 248
          FQ218531.1 - 1 250
          FQ218533.1 - 1 250
          FQ218563.1 - 1 248
          FQ218569.1 - 1 248
          FQ218570.1 - 1 250
          FQ218580.1 - 1 250
          FQ218585.1 - 1 250
          FQ218608.1 - 1 250
          FQ218609.1 - 1 250
          FQ218630.1 - 1 246
          FQ218634.1 - 1 250
          FQ218648.1 - 1 248
          FQ218657.1 - 1 250
          FQ218687.1 - 1 78
          FQ218695.1 - 1 250
          FQ218713.1 - 1 248
          FQ218744.1 - 1 232
          FQ218765.1 - 1 250
          FQ218787.1 - 1 252
          FQ218796.1 - 1 250
          FQ218800.1 - 1 250
          FQ218813.1 - 1 337
          FQ218846.1 - 1 250
          FQ218862.1 - 1 250
          FQ218872.1 - 1 250
          FQ218877.1 - 1 248
          FQ218889.1 - 1 250
          FQ218890.1 - 1 250
          FQ218892.1 - 1 250
          FQ218907.1 - 1 250
          FQ218912.1 - 1 250
          FQ218931.1 - 1 249
          FQ218948.1 - 1 250
          FQ218951.1 - 1 250
          FQ218970.1 - 1 248
          FQ218974.1 - 1 251
          FQ218980.1 - 1 250
          FQ218981.1 - 1 248
          FQ219018.1 - 1 250
          FQ219028.1 - 1 250
          FQ219033.1 - 1 250
          FQ219042.1 - 1 249
          FQ219060.1 - 1 250
          FQ219061.1 - 1 250
          FQ219086.1 - 1 250
          FQ219094.1 - 1 250
          FQ219099.1 - 1 250
          FQ219103.1 - 1 250
          FQ219110.1 - 1 250
          FQ219121.1 - 1 250
          FQ219129.1 - 1 248
          FQ219141.1 - 1 250
          FQ219164.1 - 1 250
          FQ219173.1 - 1 250
          FQ219194.1 - 1 250
          FQ219203.1 - 1 250
          FQ219221.1 - 1 250
          FQ219232.1 - 1 256
          FQ219235.1 - 1 250
          FQ219239.1 - 1 250
          FQ219251.1 - 1 250
          FQ219274.1 - 1 250
          FQ219295.1 - 1 250
          FQ219300.1 - 1 250
          FQ219311.1 - 1 250
          FQ219320.1 - 1 250
          FQ219326.1 - 1 250
          FQ219358.1 - 1 249
          FQ219363.1 - 1 252
          FQ219364.1 - 1 250
          FQ219369.1 - 1 248
          FQ219370.1 - 1 250
          FQ219377.1 - 1 250
          FQ219388.1 - 1 250
          FQ219400.1 - 1 250
          FQ219406.1 - 1 250
          FQ219412.1 - 1 250
          FQ219419.1 - 1 248
          FQ219486.1 - 1 245
          FQ219487.1 - 1 250
          FQ219488.1 - 1 250
          FQ219498.1 - 1 248
          FQ219536.1 - 1 250
          FQ219546.1 - 1 250
          FQ219551.1 - 1 250
          FQ219552.1 - 1 248
          FQ219558.1 - 1 250
          FQ219565.1 - 1 250
          FQ219587.1 - 1 248
          FQ219606.1 - 1 250
          FQ219607.1 - 1 248
          FQ219608.1 - 1 248
          FQ219617.1 - 1 250
          FQ219643.1 - 1 250
          FQ219653.1 - 1 249
          FQ219660.1 - 1 250
          FQ219663.1 - 1 250
          FQ219668.1 - 1 251
          FQ219672.1 - 1 250
          FQ219688.1 - 1 248
          FQ219691.1 - 1 250
          FQ219726.1 - 1 250
          FQ219755.1 - 1 250
          FQ219769.1 - 1 250
          FQ219770.1 - 1 250
          FQ219772.1 - 1 249
          FQ219773.1 - 1 250
          FQ219774.1 - 1 250
          FQ222367.1 - 1 248
          M29758.1 - 1 251
          X63446.1 - 1 209

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          protein complex assembly
          acute-phase response
          male gonad development
          negative regulation of endopeptidase activity
          cerebral cortex development
          negative regulation of cell growth
          negative regulation of bone mineralization
          organ regeneration
          cellular response to insulin stimulus
          negative regulation of phosphorylation
          positive regulation of bone resorption
          negative regulation of insulin receptor signaling pathway
          regulation of inflammatory response
          positive regulation of phagocytosis
          negative regulation of protein tyrosine kinase activity
          cysteine-type endopeptidase inhibitor activity
          kinase inhibitor activity
          receptor signaling protein tyrosine kinase inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0