Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPN11 SIRNA Assays ›
Gene Symbol:
Ptpn11
Gene Name:
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases:
Shp2
Chromosome Location:
Chr.12: 40895515 - 40955999 on Build Rnor_6.0
UniGene ID:
Rn.98209
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
197716
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ptpn11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001177593.1
NP_001171064.1
3
332
NM_013088.2
NP_037220.2
3
332
GenBank mRNA
U05963.1
-
3
263
U09307.1
-
3
257
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage checkpoint
activation of MAPK activity
protein dephosphorylation
lipid metabolic process
triglyceride metabolic process
epidermal growth factor receptor signaling pathway
integrin-mediated signaling pathway
axonogenesis
brain development
heart development
hormone-mediated signaling pathway
positive regulation of signal transduction
dephosphorylation
platelet formation
microvillus organization
abortive mitotic cell cycle
regulation of cell adhesion mediated by integrin
negative regulation of cell adhesion mediated by integrin
multicellular organism growth
organ growth
peptidyl-tyrosine dephosphorylation
megakaryocyte development
atrioventricular canal development
ERBB signaling pathway
regulation of multicellular organism growth
hormone metabolic process
glucose homeostasis
positive regulation of mitotic cell cycle
negative regulation of insulin secretion
regulation of protein export from nucleus
positive regulation of hormone secretion
negative regulation of hormone secretion
platelet-derived growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
ephrin receptor signaling pathway
multicellular organismal reproductive process
genitalia development
inner ear development
homeostasis of number of cells within a tissue
negative regulation of cortisol secretion
negative regulation of growth hormone secretion
face morphogenesis
intestinal epithelial cell migration
positive regulation of ERK1 and ERK2 cascade
positive regulation of glucose import in response to insulin stimulus
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
non-membrane spanning protein tyrosine phosphatase activity
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
protein domain specific binding
receptor tyrosine kinase binding
D1 dopamine receptor binding
phospholipase binding
insulin receptor substrate binding
peptide hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline