Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPSA SIRNA Assays ›
Gene Symbol:
Rpsa
Gene Name:
ribosomal protein SA
Gene Aliases:
Lamr1
Chromosome Location:
Chr.8: 128806056 - 128809987 on Build Rnor_6.0
UniGene ID:
Rn.121972
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
198122
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rpsa
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017138.2
NP_058834.1
3
262
XM_008766672.1
XP_008764894.1
3
335
GenBank mRNA
BC060578.1
-
3
280
6919235
D25224.1
-
3
263
FJ386454.1
-
2
243
FQ209463.1
-
3
276
FQ209469.1
-
3
276
FQ209490.1
-
3
277
FQ209729.1
-
3
276
FQ211189.1
-
3
276
FQ212660.1
-
3
276
FQ212893.1
-
3
276
FQ213275.1
-
3
276
FQ213317.1
-
3
276
FQ213334.1
-
3
276
FQ213623.1
-
3
279
FQ213835.1
-
3
277
FQ213894.1
-
3
276
FQ215242.1
-
3
276
FQ215292.1
-
3
277
FQ216063.1
-
3
276
FQ216085.1
-
3
276
FQ216130.1
-
3
276
FQ216245.1
-
3
276
FQ216788.1
-
3
278
FQ216997.1
-
3
276
FQ217664.1
-
3
276
FQ219586.1
-
3
276
FQ219785.1
-
3
279
FQ220012.1
-
3
276
FQ220017.1
-
3
274
FQ220075.1
-
3
277
FQ220084.1
-
3
279
FQ220126.1
-
3
276
FQ220134.1
-
3
276
FQ220171.1
-
3
275
FQ220184.1
-
3
276
FQ220347.1
-
3
278
FQ220461.1
-
3
257
FQ220468.1
-
3
276
FQ220661.1
-
3
279
FQ220822.1
-
3
276
FQ220904.1
-
3
277
FQ220964.1
-
3
276
FQ221103.1
-
3
276
FQ221334.1
-
3
296
FQ221443.1
-
3
277
FQ221672.1
-
3
276
FQ222093.1
-
3
278
FQ222280.1
-
3
276
FQ222542.1
-
3
278
FQ222757.1
-
3
274
FQ223223.1
-
3
276
FQ224747.1
-
3
276
FQ225510.1
-
3
277
FQ226054.1
-
3
276
FQ226408.1
-
3
276
FQ227056.1
-
3
276
FQ227293.1
-
3
276
FQ227600.1
-
3
277
FQ228002.1
-
3
276
FQ229290.1
-
3
278
FQ229305.1
-
3
278
FQ229367.1
-
3
277
FQ229516.1
-
3
276
FQ229579.1
-
3
276
FQ229598.1
-
3
277
FQ229712.1
-
3
276
FQ229787.1
-
3
276
FQ229850.1
-
3
278
FQ229859.1
-
3
274
FQ229890.1
-
3
276
FQ229918.1
-
3
296
FQ229945.1
-
3
276
FQ229970.1
-
3
276
FQ230088.1
-
3
276
FQ230106.1
-
3
276
FQ230180.1
-
3
277
FQ230258.1
-
3
275
FQ231469.1
-
2
93
FQ231622.1
-
3
274
FQ232305.1
-
3
278
FQ232568.1
-
3
277
FQ233068.1
-
3
276
FQ233112.1
-
3
276
FQ233340.1
-
3
278
FQ233547.1
-
3
277
FQ233919.1
-
3
276
FQ234742.1
-
3
276
FQ234765.1
-
3
276
FQ234772.1
-
3
276
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal small subunit assembly
endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
rRNA export from nucleus
translation
single organismal cell-cell adhesion
epithelial cell differentiation
structural constituent of ribosome
laminin receptor activity
structural molecule activity
ribosome binding
laminin binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0