Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACTA1 SIRNA Assays ›
          Gene Symbol:
          Acta1
          Gene Name:
          actin, alpha 1, skeletal muscle
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 56674072 - 56677084 on Build Rnor_6.0
          UniGene ID:
          Rn.82732
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 198749

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Acta1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019212.2 NP_062085.1 7 1335
          GenBank mRNA BC061974.1 - 7 1335 5623909
          FQ214585.1 - 7 1346
          FQ214598.1 - 7 1346
          FQ214600.1 - 7 1345
          FQ214603.1 - 7 1348
          FQ214610.1 - 5 889
          FQ214623.1 - 7 1343
          FQ214640.1 - 7 1344
          FQ214659.1 - 7 1346
          FQ214676.1 - 7 1345
          FQ214680.1 - 7 1345
          FQ214697.1 - 7 1350
          FQ214734.1 - 7 1345
          FQ214743.1 - 7 1344
          FQ214761.1 - 7 1343
          FQ214786.1 - 7 1347
          FQ214807.1 - 7 1344
          FQ214814.1 - 7 1344
          FQ214822.1 - 7 1345
          FQ214831.1 - 7 1344
          FQ214832.1 - 7 1343
          FQ214839.1 - 7 1347
          FQ214841.1 - 7 1344
          FQ214842.1 - 7 1345
          FQ214857.1 - 7 1348
          FQ214858.1 - 7 1344
          FQ214859.1 - 7 1344
          FQ214860.1 - 7 1346
          FQ214862.1 - 7 1344
          FQ214866.1 - 7 1346
          FQ214883.1 - 7 1343
          FQ214904.1 - 7 1344
          FQ214905.1 - 7 1345
          FQ214916.1 - 7 1344
          FQ214921.1 - 7 1345
          FQ214936.1 - 7 1345
          FQ214948.1 - 7 1344
          FQ214949.1 - 7 1345
          FQ214957.1 - 7 1344
          FQ214969.1 - 7 1344
          FQ214975.1 - 7 1347
          FQ214976.1 - 7 1344
          FQ214978.1 - 7 1344
          FQ214987.1 - 7 1344
          FQ214990.1 - 7 1344
          FQ214993.1 - 7 1344
          FQ215012.1 - 7 1345
          FQ215025.1 - 7 1346
          FQ215038.1 - 7 1343
          FQ215044.1 - 7 1344
          FQ215089.1 - 7 1345
          FQ215112.1 - 7 1344
          FQ215128.1 - 7 1346
          FQ215136.1 - 7 1344
          FQ215149.1 - 7 1346
          FQ215153.1 - 7 1344
          FQ215158.1 - 7 1342
          FQ215173.1 - 7 1345
          FQ215185.1 - 7 1344
          FQ215204.1 - 7 1342
          FQ215212.1 - 7 1346
          FQ215215.1 - 7 1345
          FQ215225.1 - 7 1345
          FQ215241.1 - 7 1345
          FQ215249.1 - 7 1345
          FQ215253.1 - 7 1345
          FQ215256.1 - 7 1344
          FQ215277.1 - 7 1345
          FQ215280.1 - 7 1345
          FQ215282.1 - 7 1344
          FQ215298.1 - 7 1345
          FQ215324.1 - 7 1344
          FQ215333.1 - 7 1344
          FQ215342.1 - 7 1344
          FQ215346.1 - 7 1344
          FQ215366.1 - 7 1345
          FQ215380.1 - 7 1346
          FQ215381.1 - 7 1345
          FQ215393.1 - 7 1344
          FQ215396.1 - 7 1344
          FQ215398.1 - 7 1344
          FQ215402.1 - 7 1344
          FQ215403.1 - 7 1345
          FQ215405.1 - 7 1343
          FQ215414.1 - 7 1346
          FQ215424.1 - 7 1345
          FQ215431.1 - 7 1345
          FQ215433.1 - 7 1343
          FQ215434.1 - 7 1345
          FQ215438.1 - 7 1344
          FQ215446.1 - 7 1344
          FQ215449.1 - 7 1346
          FQ215454.1 - 7 1345
          FQ215464.1 - 7 1344
          FQ215465.1 - 7 1344
          FQ215468.1 - 7 1344
          FQ215474.1 - 7 1345
          FQ215483.1 - 7 1344
          FQ215518.1 - 7 1353
          FQ215523.1 - 7 1344
          FQ215537.1 - 7 1346
          FQ215554.1 - 7 1344
          FQ215567.1 - 7 1346
          FQ215572.1 - 7 1345
          FQ215593.1 - 7 1344
          FQ215618.1 - 7 1345
          FQ215628.1 - 7 1347
          FQ215631.1 - 5 959
          FQ215655.1 - 7 1342
          FQ215659.1 - 7 1343
          FQ215673.1 - 7 1350
          FQ215674.1 - 7 1344
          FQ215690.1 - 7 1345
          FQ215692.1 - 7 1345
          FQ215785.1 - 7 1344
          FQ215799.1 - 7 1343
          FQ215809.1 - 7 1344
          FQ215813.1 - 7 1344
          FQ215835.1 - 7 1344
          FQ215836.1 - 7 1344
          FQ215839.1 - 7 1344
          FQ215840.1 - 7 1345
          FQ215841.1 - 7 1345
          FQ215851.1 - 7 1344
          FQ215852.1 - 7 1342
          FQ215864.1 - 7 1346
          FQ215866.1 - 7 1345
          FQ215888.1 - 7 1343
          FQ215893.1 - 7 1347
          FQ215907.1 - 7 1347
          FQ215924.1 - 5 1036
          FQ215964.1 - 7 1344
          FQ215972.1 - 7 1346
          FQ216016.1 - 7 1344
          FQ216035.1 - 7 1344
          FQ216043.1 - 7 1344
          FQ216066.1 - 7 1342
          FQ216138.1 - 7 1345
          FQ216139.1 - 7 1344
          FQ216168.1 - 7 1343
          FQ216196.1 - 7 1345
          FQ216224.1 - 7 1345
          FQ216246.1 - 7 1345
          FQ216276.1 - 7 1344
          FQ216278.1 - 7 1348
          FQ216287.1 - 7 1344
          FQ216291.1 - 7 1345
          FQ216305.1 - 7 1345
          FQ216329.1 - 7 1344
          FQ216333.1 - 7 1345
          FQ216350.1 - 7 1344
          FQ216352.1 - 7 1344
          FQ216360.1 - 7 1345
          FQ216369.1 - 7 1347
          FQ216403.1 - 7 1344
          FQ216430.1 - 7 1345
          FQ216509.1 - 7 1342
          FQ216586.1 - 7 1344
          FQ216603.1 - 7 1346
          FQ216606.1 - 7 1344
          FQ216607.1 - 7 1340
          FQ216610.1 - 7 1342
          FQ216620.1 - 7 1345
          FQ216635.1 - 7 1342
          FQ216639.1 - 7 1344
          FQ216644.1 - 7 1345
          FQ216664.1 - 7 1342
          FQ216675.1 - 7 1344
          FQ216676.1 - 7 1344
          FQ216692.1 - 7 1345
          FQ216703.1 - 7 1344
          FQ216713.1 - 7 1344
          FQ216721.1 - 7 1344
          FQ216723.1 - 7 1345
          FQ216735.1 - 3 584
          FQ216739.1 - 7 1344
          FQ216762.1 - 7 1342
          FQ216767.1 - 7 1344
          FQ216776.1 - 7 1345
          FQ216777.1 - 7 1344
          FQ216779.1 - 7 1346
          FQ216793.1 - 7 1344
          FQ216799.1 - 7 1342
          FQ216801.1 - 7 1344
          FQ216814.1 - 7 1339
          FQ216819.1 - 7 1342
          FQ216916.1 - 7 1346
          FQ216918.1 - 5 987
          FQ216961.1 - 7 1345
          FQ216994.1 - 7 1344
          FQ216999.1 - 3 543
          FQ217061.1 - 7 1346
          FQ217073.1 - 7 1347
          FQ217091.1 - 7 1343
          FQ217114.1 - 7 1346
          FQ217122.1 - 7 1344
          FQ217167.1 - 7 1344
          FQ217173.1 - 7 1344
          FQ217188.1 - 7 1344
          FQ217194.1 - 7 1344
          FQ217266.1 - 2 471
          FQ217273.1 - 7 1343
          FQ217280.1 - 7 1464
          FQ217327.1 - 7 1345
          FQ217329.1 - 7 1344
          FQ217347.1 - 7 1343
          FQ217351.1 - 2 427
          FQ217353.1 - 7 1344
          FQ217367.1 - 7 1344
          FQ217397.1 - 7 1344
          FQ217441.1 - 7 1344
          FQ217443.1 - 7 1346
          FQ217475.1 - 7 1345
          FQ217525.1 - 7 1344
          FQ217535.1 - 7 1344
          FQ217572.1 - 7 1344
          FQ217593.1 - 7 1344
          FQ217602.1 - 7 1346
          FQ217615.1 - 7 1344
          FQ217641.1 - 7 1341
          FQ217668.1 - 3 593
          FQ217762.1 - 7 1344
          FQ217826.1 - 7 1344
          FQ217855.1 - 2 419
          FQ217884.1 - 7 1343
          FQ217893.1 - 7 1345
          FQ217905.1 - 7 1344
          FQ217908.1 - 7 1346
          FQ217919.1 - 7 1344
          FQ217937.1 - 7 1344
          FQ217969.1 - 7 1346
          FQ217990.1 - 7 1344
          FQ218019.1 - 7 1345
          FQ218034.1 - 7 1344
          FQ223563.1 - 7 1345
          FQ223681.1 - 7 1348
          FQ223686.1 - 7 1344
          FQ223702.1 - 7 1345
          FQ223727.1 - 5 1042
          FQ223778.1 - 5 858
          FQ223898.1 - 7 1345
          FQ223914.1 - 7 1344
          FQ223926.1 - 7 1344
          FQ223929.1 - 7 1343
          FQ224009.1 - 7 1344
          FQ224082.1 - 7 1344
          FQ224141.1 - 7 1351
          FQ224167.1 - 7 1346
          FQ224377.1 - 7 1344
          FQ224419.1 - 7 1344
          FQ224552.1 - 7 1345
          FQ224691.1 - 7 1342
          FQ224777.1 - 7 1345
          FQ224840.1 - 7 1342
          FQ224842.1 - 7 1344
          FQ224862.1 - 7 1344
          FQ224869.1 - 7 1345
          FQ224886.1 - 7 1343
          FQ224890.1 - 7 1342
          FQ224902.1 - 7 1345
          FQ224929.1 - 7 1342
          FQ224936.1 - 7 1345
          FQ224942.1 - 7 1347
          FQ224946.1 - 5 872
          FQ224948.1 - 7 1349
          FQ224962.1 - 7 1346
          FQ224971.1 - 7 1344

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to mechanical stimulus
          response to extracellular stimulus
          response to lithium ion
          cell growth
          skeletal muscle thin filament assembly
          skeletal muscle fiber adaptation
          response to steroid hormone
          skeletal muscle fiber development
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0