Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EPB41L3 SIRNA Assays ›
          Gene Symbol:
          EPB41L3
          Gene Name:
          erythrocyte membrane protein band 4.1 like 3
          Gene Aliases:
          4.1B, DAL-1, DAL1
          Chromosome Location:
          Chr.18: 5392381 - 5630666 on Build GRCh38
          UniGene ID:
          Hs.213394
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 19971

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EPB41L3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001281533.1 NP_001268462.1 6, 7 841
          NM_001281534.1 NP_001268463.1 6, 7 1022
          NM_001281535.1 NP_001268464.1 8, 9 823
          NM_012307.3 NP_036439.2 6, 7 1022
          XM_011525623.2 XP_011523925.1 6, 7 1022
          XM_011525626.2 XP_011523928.1 5, 6 624
          XM_011525628.2 XP_011523930.1 6, 7 1022
          XM_011525632.2 XP_011523934.1 5, 6 624
          XM_011525634.2 XP_011523936.1 5, 6 624
          XM_011525637.2 XP_011523939.1 5, 6 624
          XM_017025618.1 XP_016881107.1 5, 6 624
          XM_017025619.1 XP_016881108.1 5, 6 624
          XM_017025620.1 XP_016881109.1 5, 6 624
          XM_017025621.1 XP_016881110.1 5, 6 624
          XM_017025622.1 XP_016881111.1 6, 7 1022
          XM_017025623.1 XP_016881112.1 5, 6 624
          XM_017025624.1 XP_016881113.1 5, 6 624
          XM_017025625.1 XP_016881114.1 6, 7 790
          XM_017025626.1 XP_016881115.1 5, 6 624
          XM_017025627.1 XP_016881116.1 6, 7 1022
          XM_017025628.1 XP_016881117.1 6, 7 1022
          XM_017025629.1 XP_016881118.1 6, 7 1022
          XM_017025630.1 XP_016881119.1 6, 7 790
          XM_017025631.1 XP_016881120.1 5, 6 624
          XM_017025632.1 XP_016881121.1 6, 7 1022
          XM_017025633.1 XP_016881122.1 6, 7 790
          XM_017025634.1 XP_016881123.1 5, 6 624
          XM_017025635.1 XP_016881124.1 5, 6 624
          XM_017025636.1 XP_016881125.1 6, 7 1022
          XM_017025637.1 XP_016881126.1 6, 7 1022
          XM_017025638.1 XP_016881127.1 5, 6 624
          XM_017025639.1 XP_016881128.1 6, 7 1022
          XM_017025640.1 XP_016881129.1 6, 7 1022
          XM_017025641.1 XP_016881130.1 5, 6 624
          XM_017025642.1 XP_016881131.1 6, 7 1022
          XM_017025643.1 XP_016881132.1 6, 7 1022
          XM_017025644.1 XP_016881133.1 6, 7 1022
          XM_017025645.1 XP_016881134.1 6, 7 881
          XM_017025646.1 XP_016881135.1 6, 7 1022
          XM_017025647.1 XP_016881136.1 6, 7 881
          XM_017025648.1 XP_016881137.1 6, 7 881
          XM_017025649.1 XP_016881138.1 6, 7 881
          XM_017025650.1 XP_016881139.1 6, 7 881
          XM_017025651.1 XP_016881140.1 6, 7 881
          XM_017025652.1 XP_016881141.1 6, 7 881
          XM_017025653.1 XP_016881142.1 6, 7 881
          XM_017025654.1 XP_016881143.1 6, 7 881
          XM_017025655.1 XP_016881144.1 6, 7 881
          XM_017025656.1 XP_016881145.1 6, 7 881
          XM_017025657.1 XP_016881146.1 6, 7 881
          XM_017025658.1 XP_016881147.1 6, 7 881
          XM_017025659.1 XP_016881148.1 6, 7 881
          XM_017025660.1 XP_016881149.1 6, 7 881
          GenBank mRNA AB023204.1 - 6, 7 699
          AF069072.1 - 6, 7 679
          AK128531.1 - 7, 8 690
          AK292583.1 - 8, 9 823
          AK297406.1 - 6, 7 841
          AK307904.1 - 6, 7 943
          AK314438.1 - 4, 5 349
          BC006141.1 - 6, 7 993 3936453
          BC008377.1 - 6, 7 858

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053927, XM_006245651, XM_006245649, XM_006245652, XM_008767417, and others.
          This siRNA also targets the following transcripts in Mouse: NM_013813, AF152247, AF177146, HG738870, AK197868, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          protein localization to paranode region of axon
          apoptotic process
          cytoskeletal anchoring at plasma membrane
          biological_process
          regulation of cell shape
          cortical cytoskeleton organization
          cortical actin cytoskeleton organization
          paranodal junction assembly
          myelin maintenance
          neuron projection morphogenesis
          protein localization to juxtaparanode region of axon
          protein localization to plasma membrane
          actin binding
          structural constituent of cytoskeleton
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0