Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BDNF SIRNA Assays ›
          Gene Symbol:
          Bdnf
          Gene Name:
          brain-derived neurotrophic factor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 100768637 - 100819216 on Build Rnor_6.0
          UniGene ID:
          Rn.11266
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 199801

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Bdnf
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270630.1 NP_001257559.1 2 799
          NM_001270631.1 NP_001257560.1 2 950
          NM_001270632.1 NP_001257561.1 2 867
          NM_001270633.1 NP_001257562.1 2 791
          NM_001270634.1 NP_001257563.1 3 678
          NM_001270635.1 NP_001257564.1 2 677
          NM_001270636.1 NP_001257565.1 2 663
          NM_001270637.1 NP_001257566.1 2 655
          NM_001270638.1 NP_001257567.1 2 532
          NM_012513.4 NP_036645.2 2 1090
          XM_006234684.2 XP_006234746.1 2 798
          XM_008762078.1 XP_008760300.1 2 2724
          GenBank mRNA AY176065.1 - 1 430
          AY559248.1 - 2 593
          AY559249.1 - 2 607
          AY559250.1 - 3 679
          BC087634.1 - 2 785 7319966
          EF125675.1 - 2 1090
          EF125676.1 - 2 656
          EF125677.1 - 2 868
          EF125678.1 - 2 951
          EF125679.1 - 2 791
          EF125680.1 - 2 799
          EF125686.1 - 2 677
          EF125687.1 - 2 532
          EF125688.1 - 2 664
          EF125689.1 - 2 728
          EF125690.1 - 1 922
          M61175.1 - 2 502
          X67108.1 - 1 451

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001143806, NM_001143810, NM_001143811, NM_001143805, NM_001143807, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001285420, NM_001285419, NM_001285417, NM_001285421, NM_001048139, and others.

          Panther Classification:

          Molecular Function -

          neurotrophic factor

          Gene Ontology Categories:

          Function(s) Process(es)

          ureteric bud development
          behavioral fear response
          response to hypoxia
          chronic inflammatory response
          mitochondrial electron transport, NADH to ubiquinone
          transmembrane receptor protein tyrosine kinase signaling pathway
          gamma-aminobutyric acid signaling pathway
          nervous system development
          negative regulation of neuroblast proliferation
          axon guidance
          axon target recognition
          learning or memory
          learning
          circadian rhythm
          feeding behavior
          neuron recognition
          response to light stimulus
          response to light intensity
          response to hormone
          response to auditory stimulus
          glutamate secretion
          response to fluoxetine
          response to activity
          dendrite development
          regulation of metabolic process
          nerve development
          neuron differentiation
          regulation of axon extension
          response to nutrient levels
          response to food
          negative regulation of synaptic transmission, GABAergic
          positive regulation of peptidyl-serine phosphorylation
          response to vitamin A
          response to tumor necrosis factor
          response to potassium ion
          mechanoreceptor differentiation
          response to drug
          fear response
          negative regulation of apoptotic process
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of neuron differentiation
          negative regulation of striated muscle tissue development
          regulation of retinal cell programmed cell death
          regulation of synaptic plasticity
          regulation of long-term neuronal synaptic plasticity
          positive regulation of long-term neuronal synaptic plasticity
          regulation of short-term neuronal synaptic plasticity
          axon extension
          inner ear development
          cognition
          positive regulation of sequence-specific DNA binding transcription factor activity
          response to electrical stimulus
          positive regulation of synapse assembly
          response to hyperoxia
          excitatory postsynaptic potential
          inhibitory postsynaptic potential
          negative regulation of cell death
          taste bud development
          cellular response to tumor necrosis factor
          cellular response to norepinephrine stimulus
          response to anesthetic
          dendrite extension
          negative regulation of neuron death
          response to L-glutamate
          response to nerve growth factor
          cellular response to nerve growth factor stimulus
          neuron projection extension
          positive regulation of glucocorticoid receptor signaling pathway
          neurotrophin TRKB receptor binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0