Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAT SIRNA Assays ›
Gene Symbol:
Cat
Gene Name:
catalase
Gene Aliases:
CS1, Cas1, Cat01, Catl, Cs-1
Chromosome Location:
Chr.3: 93379872 - 93412058 on Build Rnor_6.0
UniGene ID:
Rn.3001
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
199818
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Cat
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012520.2
NP_036652.1
2
239
GenBank mRNA
BC081853.1
-
2
136
7109889
FM043772.1
-
2
239
M11670.1
-
2
190
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009804, AK133881, AY040626, AK159891, AK148961, and others.
Panther Classification:
Molecular Function -
peroxidase
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
osteoblast differentiation
ureteric bud development
response to hypoxia
kidney development
triglyceride metabolic process
response to oxidative stress
aging
cholesterol metabolic process
aerobic respiration
response to radiation
response to UV
response to toxic substance
response to light intensity
UV protection
response to ozone
response to lead ion
positive regulation of phosphatidylinositol 3-kinase signaling
response to activity
response to inactivity
hemoglobin metabolic process
negative regulation of NF-kappaB transcription factor activity
response to estradiol
response to insulin
response to vitamin A
response to vitamin E
response to L-ascorbic acid
response to drug
hydrogen peroxide catabolic process
negative regulation of apoptotic process
response to ethanol
response to cadmium ion
positive regulation of NF-kappaB transcription factor activity
protein tetramerization
protein homotetramerization
positive regulation of cell division
response to hyperoxia
oxidation-reduction process
response to fatty acid
cellular response to growth factor stimulus
response to phenylpropanoid
aminoacylase activity
catalase activity
receptor binding
antioxidant activity
oxidoreductase activity, acting on peroxide as acceptor
enzyme binding
heme binding
protein homodimerization activity
metal ion binding
NADP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-bznxf:80/100.66.77.153:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline