Search
Search

Dmd| Gene Symbol: Dmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005246.1 | NP_001005246.1 | 4 | 386 | ||
| XM_008773252.1 | XP_008771474.1 | 4 | 453 | |||
| XM_008773253.1 | XP_008771475.1 | 4 | 453 | |||
| GenBank mRNA | X69767.1 | - | 4 | 401 | ||