Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APOA1 SIRNA Assays ›
          Gene Symbol:
          Apoa1
          Gene Name:
          apolipoprotein A-I
          Gene Aliases:
          apoA-I
          Chromosome Location:
          Chr.8: 50525091 - 50526875 on Build Rnor_6.0
          UniGene ID:
          Rn.10308
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 200325

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Apoa1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012738.1 NP_036870.1 4 495
          GenBank mRNA BC089820.1 - 3 479 7321674
          FQ209434.1 - 4 496
          FQ209676.1 - 4 500
          FQ209742.1 - 4 500
          FQ209761.1 - 4 496
          FQ209776.1 - 5 451
          FQ209818.1 - 4 501
          FQ209900.1 - 4 496
          FQ209928.1 - 4 500
          FQ209958.1 - 4 500
          FQ210010.1 - 4 500
          FQ210126.1 - 4 497
          FQ210161.1 - 4 523
          FQ210244.1 - 1 157
          FQ210354.1 - 4 500
          FQ210367.1 - 4 496
          FQ210371.1 - 4 496
          FQ210470.1 - 4 496
          FQ210544.1 - 4 498
          FQ210653.1 - 4 498
          FQ210722.1 - 4 500
          FQ210732.1 - 1 137
          FQ210743.1 - 4 603
          FQ210880.1 - 4 496
          FQ210914.1 - 4 496
          FQ210920.1 - 4 496
          FQ211297.1 - 4 498
          FQ211535.1 - 4 503
          FQ211571.1 - 4 496
          FQ218078.1 - 4 496
          FQ218079.1 - 4 500
          FQ218325.1 - 4 500
          FQ218519.1 - 4 496
          FQ218525.1 - 4 496
          FQ218615.1 - 4 495
          FQ218659.1 - 4 499
          FQ218837.1 - 4 502
          FQ219021.1 - 4 500
          FQ219052.1 - 4 502
          FQ219074.1 - 4 496
          FQ219479.1 - 4 588
          FQ219515.1 - 4 500
          FQ219577.1 - 4 496
          FQ219728.1 - 4 495
          FQ224655.1 - 4 496
          FQ229884.1 - 4 535
          M00001.1 - 4 495
          X00558.1 - 3 484

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein phosphorylation
          endothelial cell proliferation
          negative regulation of cytokine secretion involved in immune response
          phospholipid metabolic process
          phosphatidylcholine biosynthetic process
          cholesterol biosynthetic process
          transforming growth factor beta receptor signaling pathway
          G-protein coupled receptor signaling pathway
          integrin-mediated signaling pathway
          response to nutrient
          protein localization
          cholesterol metabolic process
          glucocorticoid metabolic process
          negative regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of cholesterol esterification
          positive regulation of triglyceride catabolic process
          negative regulation of very-low-density lipoprotein particle remodeling
          peripheral nervous system axon regeneration
          phospholipid transport
          protein oxidation
          peptidyl-methionine modification
          lipid storage
          regulation of intestinal cholesterol absorption
          cholesterol transport
          adrenal gland development
          organ regeneration
          axon regeneration
          regulation of Cdc42 protein signal transduction
          cholesterol efflux
          phospholipid efflux
          negative regulation of heterotypic cell-cell adhesion
          very-low-density lipoprotein particle remodeling
          high-density lipoprotein particle assembly
          positive regulation of Rho protein signal transduction
          lipoprotein metabolic process
          lipoprotein biosynthetic process
          response to drug
          cholesterol homeostasis
          blood vessel endothelial cell migration
          response to estrogen
          reverse cholesterol transport
          positive regulation of fatty acid biosynthetic process
          phosphatidylcholine metabolic process
          negative regulation of interleukin-1 beta secretion
          negative regulation of inflammatory response
          protein stabilization
          negative chemotaxis
          positive regulation of lipoprotein lipase activity
          vitamin transport
          positive regulation of hydrolase activity
          negative regulation of hydrolase activity
          positive regulation of transferase activity
          positive regulation of stress fiber assembly
          phospholipid homeostasis
          negative regulation of lipase activity
          negative regulation of cell adhesion molecule production
          negative regulation of response to cytokine stimulus
          triglyceride homeostasis
          ERK1 and ERK2 cascade
          cholesterol import
          positive regulation of substrate adhesion-dependent cell spreading
          beta-amyloid binding
          lipid transporter activity
          phospholipid binding
          phospholipid transporter activity
          high-density lipoprotein particle binding
          lipid binding
          cholesterol binding
          cholesterol transporter activity
          enzyme binding
          phosphatidylcholine binding
          apolipoprotein receptor binding
          apolipoprotein A-I receptor binding
          identical protein binding
          chemorepellent activity
          lipase inhibitor activity
          phosphatidylcholine-sterol O-acyltransferase activator activity
          high-density lipoprotein particle receptor binding
          lipoprotein particle binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline