Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AQP1 SIRNA Assays ›
Gene Symbol:
Aqp1
Gene Name:
aquaporin 1
Gene Aliases:
CHIP28
Chromosome Location:
Chr.4: 85551503 - 85563683 on Build Rnor_6.0
UniGene ID:
Rn.1618
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200419
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Aqp1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012778.1
NP_036910.1
4
1780
GenBank mRNA
BC090068.1
-
4
1762
7190406
X67948.1
-
4
1780
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
Gene Ontology Categories:
Function(s)
Process(es)
glomerular filtration
renal water transport
cGMP biosynthetic process
potassium ion transport
water transport
cell volume homeostasis
hyperosmotic response
response to hormone
cellular water homeostasis
positive regulation of lamellipodium assembly
positive regulation of epithelial cell migration
carbon dioxide transport
ammonium transport
glycerol transport
sensory perception of pain
cellular homeostasis
lateral ventricle development
water homeostasis
pancreatic juice secretion
nitric oxide transport
positive regulation of cell migration
establishment or maintenance of actin cytoskeleton polarity
secretion by cell
cerebrospinal fluid secretion
secretory granule organization
cellular response to UV
transepithelial water transport
carbon dioxide transmembrane transport
wound healing
odontogenesis
response to drug
hyperosmotic salinity response
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to estrogen
lipid digestion
positive regulation of angiogenesis
positive regulation of saliva secretion
positive regulation of fibroblast proliferation
camera-type eye morphogenesis
multicellular organismal water homeostasis
corticotropin secretion
renal water absorption
cellular response to hydrogen peroxide
cellular response to inorganic substance
cellular response to mechanical stimulus
cellular response to copper ion
cellular response to mercury ion
cellular response to retinoic acid
cellular response to cAMP
cellular response to hypoxia
cellular response to salt stress
cellular hyperosmotic response
cellular response to dexamethasone stimulus
cellular response to nitric oxide
potassium ion transmembrane transport
metanephric descending thin limb development
metanephric proximal straight tubule development
metanephric proximal convoluted tubule segment 2 development
metanephric glomerulus vasculature development
ammonium transmembrane transport
maintenance of symbiont-containing vacuole by host
cation transmembrane transport
intracellular cGMP activated cation channel activity
potassium channel activity
water transmembrane transporter activity
protein binding
ammonium transmembrane transporter activity
potassium ion transmembrane transporter activity
glycerol transmembrane transporter activity
water channel activity
glycerol channel activity
transmembrane transporter activity
nitric oxide transmembrane transporter activity
carbon dioxide transmembrane transporter activity
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0