Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPO SIRNA Assays ›
Gene Symbol:
Epo
Gene Name:
erythropoietin
Gene Aliases:
-
Chromosome Location:
Chr.12: 22274828 - 22278268 on Build Rnor_6.0
UniGene ID:
Rn.11365
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200564
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Epo
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017001.1
NP_058697.1
4
285
XM_006249101.2
XP_006249163.1
3
427
GenBank mRNA
D10763.1
-
4
285
L10608.1
-
3
254
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
regulation of transcription from RNA polymerase II promoter
apoptotic process
embryo implantation
aging
response to nutrient
positive regulation of cell proliferation
response to salt stress
negative regulation of calcium ion transport into cytosol
peptidyl-serine phosphorylation
erythrocyte differentiation
response to lipopolysaccharide
negative regulation of myeloid cell apoptotic process
response to vitamin A
response to testosterone
positive regulation of tyrosine phosphorylation of Stat5 protein
hemoglobin biosynthetic process
negative regulation of apoptotic process
erythrocyte maturation
response to estrogen
positive regulation of neuron differentiation
positive regulation of DNA replication
positive regulation of protein kinase activity
positive regulation of transcription, DNA-templated
positive regulation of Ras protein signal transduction
response to axon injury
response to electrical stimulus
response to hyperoxia
response to interleukin-1
cellular hyperosmotic response
negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress
negative regulation of erythrocyte apoptotic process
negative regulation of cation channel activity
erythropoietin receptor binding
hormone activity
protein kinase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0