Search
Search

Gfap| Gene Symbol: Gfap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017009.2 | NP_058705.2 | 5 | 840 | ||
| XM_008768261.1 | XP_008766483.1 | 5 | 906 | |||
| GenBank mRNA | BC088851.1 | - | 5 | 840 | 7315338 | |
| EF094476.1 | - | 5 | 911 | |||
| L27219.1 | - | 5 | 844 | |||
| U03700.1 | - | 5 | 864 | |||