Search
Search

Idh1| Gene Symbol: Idh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031510.1 | NP_113698.1 | 1 | 124 | ||
| XM_006245049.2 | XP_006245111.1 | 2 | 182 | |||
| XM_008767076.1 | XP_008765298.1 | 2 | 214 | |||
| GenBank mRNA | DQ403076.1 | - | 1 | 14 | ||
| FQ214242.1 | - | 2 | 205 | |||
| L35317.1 | - | 1 | 124 | |||