Search
Search

Fmr1| Gene Symbol: Fmr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052804.1 | NP_434691.1 | 3 | 343 | ||
| XM_006229530.2 | XP_006229592.1 | 3 | 393 | |||
| XM_006229531.2 | XP_006229593.1 | 3 | 393 | |||
| XM_006229532.2 | XP_006229594.1 | 3 | 393 | |||
| XM_006229535.2 | XP_006229597.1 | 2 | 112 | |||
| XM_008773597.1 | XP_008771819.1 | 3 | 225 | |||
| GenBank mRNA | AF435434.1 | - | 3 | 162 | ||
| AY240947.1 | - | 3 | 343 | |||
| U60145.1 | - | 1 | 52 | |||