Search
Search

MEP1B| Gene Symbol: MEP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308171.1 | NP_001295100.1 | 7, 8 | 602 | ||
| NM_005925.2 | NP_005916.2 | 7, 8 | 602 | |||
| GenBank mRNA | AY695931.1 | - | 7, 8 | 613 | ||
| BC136559.1 | - | 7, 8 | 580 | 9020549 | ||
| BC144244.1 | - | 7, 8 | 580 | 9052764 | ||
| X81333.1 | - | 7, 8 | 601 | |||