Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EZH2 SIRNA Assays ›
          Gene Symbol:
          EZH2
          Gene Name:
          enhancer of zeste 2 polycomb repressive complex 2 subunit
          Gene Aliases:
          ENX-1, ENX1, EZH1, EZH2b, KMT6, KMT6A, WVS, WVS2
          Chromosome Location:
          Chr.7: 148807372 - 148884662 on Build GRCh38
          UniGene ID:
          Hs.444082
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 213529

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EZH2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001203247.1 NP_001190176.1 7 877
          NM_001203248.1 NP_001190177.1 7 850
          NM_001203249.1 NP_001190178.1 7 977
          NM_004456.4 NP_004447.2 7 877
          NM_152998.2 NP_694543.1 6 760
          XM_005249962.4 XP_005250019.1 7 3318
          XM_005249963.4 XP_005250020.1 7 3291
          XM_005249964.4 XP_005250021.1 7 3292
          XM_011515883.2 XP_011514185.1 8 852
          XM_011515884.2 XP_011514186.1 7 984
          XM_011515885.2 XP_011514187.1 7 3293
          XM_011515886.2 XP_011514188.1 8 1001
          XM_011515887.2 XP_011514189.1 7 850
          XM_011515888.2 XP_011514190.1 7 972
          XM_011515889.2 XP_011514191.1 7 774
          XM_011515890.2 XP_011514192.1 6 3203
          XM_011515891.2 XP_011514193.1 5 937
          XM_011515892.2 XP_011514194.1 7 3321
          XM_011515893.2 XP_011514195.1 6 3201
          XM_011515894.2 XP_011514196.1 6 702
          XM_011515895.2 XP_011514197.1 6 3176
          XM_011515896.2 XP_011514198.1 6 3202
          XM_011515897.2 XP_011514199.1 8 1028
          XM_011515898.2 XP_011514200.1 8 955
          XM_011515899.2 XP_011514201.1 7 3323
          XM_011515901.2 XP_011514203.1 7 3323
          XM_017011817.1 XP_016867306.1 7 3319
          XM_017011818.1 XP_016867307.1 8 1001
          XM_017011819.1 XP_016867308.1 7 3319
          XM_017011820.1 XP_016867309.1 6 733
          XM_017011821.1 XP_016867310.1 8 1028
          GenBank mRNA AB208895.1 - 7 811
          AK092676.1 - 6 659
          AK293239.1 - 7 788
          AK302216.1 - 7 977
          AK303585.1 - 7 815
          AK303637.1 - 8 1029
          AK314291.1 - 6 702
          AY519465.1 - 6 684
          BC010858.2 - 7 806 3901250
          JF327843.1 - 5 567
          U61145.1 - 7 773

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001134979, XM_006236404, XM_006236408, XM_006236403, XM_006236405, and others.
          This siRNA also targets the following transcripts in Mouse: NM_007971, NM_001146689, BC016391, BC003772, U52951, and others.

          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          DNA methylation
          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          positive regulation of epithelial to mesenchymal transition
          regulation of gliogenesis
          skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
          cerebellar cortex development
          hippocampus development
          response to estradiol
          negative regulation of transcription elongation from RNA polymerase II promoter
          regulation of cell proliferation
          regulation of circadian rhythm
          positive regulation of MAP kinase activity
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of GTPase activity
          negative regulation of epidermal cell differentiation
          negative regulation of gene expression, epigenetic
          negative regulation of transcription, DNA-templated
          negative regulation of retinoic acid receptor signaling pathway
          rhythmic process
          negative regulation of striated muscle cell differentiation
          cellular response to hydrogen peroxide
          G1 to G0 transition
          histone H3-K27 methylation
          protein localization to chromatin
          positive regulation of protein serine/threonine kinase activity
          positive regulation of dendrite development
          negative regulation of G1/S transition of mitotic cell cycle
          core promoter binding
          DNA binding
          chromatin binding
          RNA binding
          protein binding
          protein-lysine N-methyltransferase activity
          histone-lysine N-methyltransferase activity
          chromatin DNA binding
          histone methyltransferase activity
          ribonucleoprotein complex binding
          sequence-specific DNA binding
          histone methyltransferase activity (H3-K27 specific)
          promoter-specific chromatin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0