Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EZH2 SIRNA Assays ›
          Gene Symbol:
          EZH2
          Gene Name:
          enhancer of zeste 2 polycomb repressive complex 2 subunit
          Gene Aliases:
          ENX-1, ENX1, EZH1, EZH2b, KMT6, KMT6A, WVS, WVS2
          Chromosome Location:
          Chr.7: 148807372 - 148884662 on Build GRCh38
          UniGene ID:
          Hs.444082
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 214021

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EZH2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001203247.1 NP_001190176.1 20 2391
          NM_001203248.1 NP_001190177.1 20 2364
          NM_001203249.1 NP_001190178.1 19 2365
          NM_004456.4 NP_004447.2 20 2406
          NM_152998.2 NP_694543.1 19 2274
          XM_005249962.4 XP_005250019.1 20 4832
          XM_005249963.4 XP_005250020.1 20 4805
          XM_005249964.4 XP_005250021.1 19 4680
          XM_011515883.2 XP_011514185.1 21 2381
          XM_011515884.2 XP_011514186.1 20 2513
          XM_011515885.2 XP_011514187.1 20 4822
          XM_011515886.2 XP_011514188.1 21 2530
          XM_011515887.2 XP_011514189.1 20 2379
          XM_011515888.2 XP_011514190.1 20 2501
          XM_011515889.2 XP_011514191.1 20 2303
          XM_011515890.2 XP_011514192.1 19 4732
          XM_011515891.2 XP_011514193.1 18 2466
          XM_011515892.2 XP_011514194.1 19 4724
          XM_011515893.2 XP_011514195.1 19 4715
          XM_011515894.2 XP_011514196.1 19 2231
          XM_011515895.2 XP_011514197.1 19 4705
          XM_011515896.2 XP_011514198.1 18 4590
          XM_011515897.2 XP_011514199.1 21 2557
          XM_011515898.2 XP_011514200.1 21 2484
          XM_017011817.1 XP_016867306.1 20 4848
          XM_017011818.1 XP_016867307.1 21 2515
          XM_017011819.1 XP_016867308.1 19 4707
          XM_017011820.1 XP_016867309.1 19 2262
          XM_017011821.1 XP_016867310.1 21 2542
          GenBank mRNA AB208895.1 - 19 3335
          AK092676.1 - 19 2173
          AK293239.1 - 20 2302
          AK302216.1 - 19 2365
          AK314291.1 - 19 2216
          BC010858.2 - 20 2335 3901250
          BU953788.1 - 3 187
          JF327843.1 - 17 1955
          U61145.1 - 20 2287
          X95653.1 - 20 2255

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001134979, XM_006236404, XM_006236408, XM_006236403, XM_006236405, and others.
          This siRNA also targets the following transcripts in Mouse: NM_007971, NM_001146689, BC016391, BC003772, U52951, and others.

          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          DNA methylation
          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          positive regulation of epithelial to mesenchymal transition
          regulation of gliogenesis
          skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
          cerebellar cortex development
          hippocampus development
          response to estradiol
          negative regulation of transcription elongation from RNA polymerase II promoter
          regulation of cell proliferation
          regulation of circadian rhythm
          positive regulation of MAP kinase activity
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of GTPase activity
          negative regulation of epidermal cell differentiation
          negative regulation of gene expression, epigenetic
          negative regulation of transcription, DNA-templated
          negative regulation of retinoic acid receptor signaling pathway
          rhythmic process
          negative regulation of striated muscle cell differentiation
          cellular response to hydrogen peroxide
          G1 to G0 transition
          histone H3-K27 methylation
          protein localization to chromatin
          positive regulation of protein serine/threonine kinase activity
          positive regulation of dendrite development
          negative regulation of G1/S transition of mitotic cell cycle
          core promoter binding
          DNA binding
          chromatin binding
          RNA binding
          protein binding
          protein-lysine N-methyltransferase activity
          histone-lysine N-methyltransferase activity
          chromatin DNA binding
          histone methyltransferase activity
          ribonucleoprotein complex binding
          sequence-specific DNA binding
          histone methyltransferase activity (H3-K27 specific)
          promoter-specific chromatin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0