Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCL2 SIRNA Assays ›
          Gene Symbol:
          BCL2
          Gene Name:
          BCL2, apoptosis regulator
          Gene Aliases:
          Bcl-2, PPP1R50
          Chromosome Location:
          Chr.18: 63123346 - 63319778 on Build GRCh38
          UniGene ID:
          Hs.150749
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 214533

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BCL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000633.2 NP_000624.2 2 551
          NM_000657.2 NP_000648.2 2 551
          XM_011526135.2 XP_011524437.1 1 1170
          XM_017025917.1 XP_016881406.1 2 949
          GenBank mRNA BC027258.1 - 2 447 4511027
          EU287875.1 - 1 58
          M13994.1 - 1 1516
          M13995.1 - 1 204
          M14745.1 - 1 89
          X06487.1 - 2 944

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_016993, U34964, L14680.

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          ossification
          ovarian follicle development
          metanephros development
          branching involved in ureteric bud morphogenesis
          behavioral fear response
          B cell homeostasis
          release of cytochrome c from mitochondria
          regulation of cell-matrix adhesion
          lymphoid progenitor cell differentiation
          B cell lineage commitment
          response to ischemia
          renal system process
          protein dephosphorylation
          melanin metabolic process
          regulation of nitrogen utilization
          apoptotic process
          humoral immune response
          cellular response to DNA damage stimulus
          actin filament organization
          axonogenesis
          female pregnancy
          cell aging
          male gonad development
          extrinsic apoptotic signaling pathway via death domain receptors
          intrinsic apoptotic signaling pathway in response to DNA damage
          intrinsic apoptotic signaling pathway in response to oxidative stress
          response to radiation
          response to toxic substance
          post-embryonic development
          response to iron ion
          response to UV-B
          response to gamma radiation
          regulation of gene expression
          negative regulation of autophagy
          negative regulation of calcium ion transport into cytosol
          regulation of glycoprotein biosynthetic process
          mesenchymal cell development
          positive regulation of neuron maturation
          positive regulation of smooth muscle cell migration
          cell growth
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          cochlear nucleus development
          gland morphogenesis
          regulation of transmembrane transporter activity
          negative regulation of ossification
          positive regulation of cell growth
          negative regulation of cell growth
          melanocyte differentiation
          negative regulation of cell migration
          positive regulation of B cell proliferation
          hair follicle morphogenesis
          axon regeneration
          regulation of protein stability
          endoplasmic reticulum calcium ion homeostasis
          glomerulus development
          negative regulation of cellular pH reduction
          negative regulation of myeloid cell apoptotic process
          T cell differentiation in thymus
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of osteoblast proliferation
          response to cytokine
          response to nicotine
          organ growth
          positive regulation of multicellular organism growth
          B cell proliferation
          cellular response to glucose starvation
          response to drug
          response to hydrogen peroxide
          T cell homeostasis
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          CD8-positive, alpha-beta T cell lineage commitment
          regulation of protein homodimerization activity
          regulation of protein heterodimerization activity
          negative regulation of neuron apoptotic process
          ear development
          regulation of viral genome replication
          positive regulation of melanocyte differentiation
          negative regulation of retinal cell programmed cell death
          regulation of mitochondrial membrane permeability
          focal adhesion assembly
          spleen development
          thymus development
          digestive tract morphogenesis
          oocyte development
          positive regulation of skeletal muscle fiber development
          pigment granule organization
          homeostasis of number of cells within a tissue
          B cell receptor signaling pathway
          response to glucocorticoid
          neuron apoptotic process
          defense response to virus
          regulation of mitochondrial membrane potential
          negative regulation of mitochondrial depolarization
          regulation of calcium ion transport
          transmembrane transport
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to organic substance
          cellular response to hypoxia
          reactive oxygen species metabolic process
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          negative regulation of G1/S transition of mitotic cell cycle
          negative regulation of reactive oxygen species metabolic process
          negative regulation of anoikis
          negative regulation of apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          negative regulation of intrinsic apoptotic signaling pathway
          positive regulation of intrinsic apoptotic signaling pathway
          protease binding
          protein binding
          transcription factor binding
          channel activity
          channel inhibitor activity
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity
          sequence-specific DNA binding
          protein heterodimerization activity
          BH3 domain binding
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0