Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PGAP2 SIRNA Assays ›
Gene Symbol:
PGAP2
Gene Name:
post-GPI attachment to proteins 2
Gene Aliases:
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
Chromosome Location:
Chr.11: 3797724 - 3826371 on Build GRCh38
UniGene ID:
Hs.133968
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21495
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PGAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145438.2
NP_001138910.1
4, 5
615
NM_001256235.1
NP_001243164.1
5, 6
556
NM_001256236.1
NP_001243165.1
5, 6
798
NM_001256237.1
NP_001243166.1
4, 5
615
NM_001256238.1
NP_001243167.1
4, 5
529
NM_001256239.1
NP_001243168.1
3, 4
545
NM_001256240.1
NP_001243169.1
3, 4
545
NM_001283038.1
NP_001269967.1
4, 5
615
NM_001283039.1
NP_001269968.1
4, 5
568
NM_014489.3
NP_055304.1
4, 5
728
NR_027016.2
2, 3
370
NR_027017.2
5, 6
678
NR_027018.2
5, 6
567
NR_045923.1
3, 4
440
NR_045927.1
3, 4
440
NR_045929.1
3, 4
498
NR_104270.1
3, 4
440
NR_104271.1
5, 6
700
NR_104272.1
2, 3
370
XM_006718181.3
XP_006718244.1
3, 4
1206
XM_006718183.3
XP_006718246.1
5, 6
623
XM_006718185.2
XP_006718248.1
4, 5
515
XM_006718186.2
XP_006718249.1
5, 6
675
XM_006718190.3
XP_006718253.1
4, 5
831
XM_006718191.3
XP_006718254.1
4, 5
831
XM_006718192.1
XP_006718255.1
6, 7
749
XM_006718193.2
XP_006718256.1
5, 6
556
XM_011519990.2
XP_011518292.2
5, 6
1082
XM_011519991.2
XP_011518293.2
5, 6
1085
XM_011519992.1
XP_011518294.1
6, 7
854
XM_011519993.2
XP_011518295.2
5, 6
1101
XM_011519994.2
XP_011518296.2
5, 6
1101
XM_011519995.2
XP_011518297.1
5, 6
1005
XM_011519996.1
XP_011518298.1
6, 7
806
XM_011519997.1
XP_011518299.1
6, 7
1147
XM_011519998.2
XP_011518300.1
6, 7
825
XM_011519999.1
XP_011518301.1
6, 7
784
XM_011520000.1
XP_011518302.1
5, 6
698
XM_011520001.1
XP_011518303.1
4, 5
490
XM_011520002.1
XP_011518304.1
4, 5
490
XM_011520003.2
XP_011518305.1
5, 6
1014
XM_011520004.2
XP_011518306.1
5, 6
689
XM_011520005.2
XP_011518307.1
4, 5
586
XM_011520006.2
XP_011518308.1
5, 6
970
XM_011520007.2
XP_011518309.1
6, 7
739
XM_017017556.1
XP_016873045.1
4, 5
490
XM_017017557.1
XP_016873046.1
4, 5
638
XM_017017558.1
XP_016873047.1
4, 5
812
XM_017017559.1
XP_016873048.1
5, 6
675
XM_017017560.1
XP_016873049.1
5, 6
675
GenBank mRNA
AF159615.1
-
4, 5
730
AK292181.1
-
5, 6
692
AK295202.1
-
5, 6
567
AK301991.1
-
5, 6
648
AK307359.1
-
3, 4
357
AL096753.1
-
5, 6
556
AY373030.1
-
2, 3
419
BC009930.2
-
4, 5
502
3506614
BG707484.1
-
3, 4
334
BI759334.1
-
3, 4
437
BI768071.1
-
3, 4
502
BI771208.1
-
2, 3
331
BI772237.1
-
4, 5
508
BI911526.1
-
5, 6
589
BX340489.2
-
3, 4
482
BX356273.2
-
4, 5
522
BX379511.2
-
4, 5
497
BX448155.2
-
4, 5
474
CB988453.1
-
2, 3
397
CD675009.1
-
1, 2
81
CD675113.1
-
3, 4
333
CX872317.1
-
2, 3
298
KU178608.1
-
2, 3
419
KU178609.1
-
3, 4
504
KU178610.1
-
3, 4
504
KU178611.1
-
2, 3
419
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001291358, NM_145583, AK159933, AK146442, BC004794, and others.
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
protein transport
signal transduction in response to DNA damage
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fz59j:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0