Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS3 SIRNA Assays ›
          Gene Symbol:
          RPS3
          Gene Name:
          ribosomal protein S3
          Gene Aliases:
          S3
          Chromosome Location:
          Chr.11: 75399491 - 75422302 on Build GRCh38
          UniGene ID:
          Hs.546286
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 215079

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RPS3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005.4 NP_000996.2 7 814
          NM_001260506.1 NP_001247435.1 7 893
          NM_001260507.1 NP_001247436.1 6 683
          GenBank mRNA AB062288.1 - 7 750
          AK129794.1 - 7 693
          AK130960.1 - 7 637
          AK223048.1 - 7 763
          BC003137.1 - 7 763 2906158
          BC003577.1 - 7 755
          BC013196.1 - 7 755
          BC013231.1 - 7 775
          BC029981.1 - 6 708
          BC034149.1 - 7 787 4665438
          BC071917.1 - 7 766 6297012
          BC100284.1 - 7 775 6160514
          S42658.1 - 7 765

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          DNA repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          rRNA processing
          translation
          translational initiation
          SRP-dependent cotranslational protein targeting to membrane
          apoptotic process
          cellular response to DNA damage stimulus
          response to oxidative stress
          chromosome segregation
          mitotic nuclear division
          positive regulation of gene expression
          negative regulation of translation
          viral transcription
          positive regulation of microtubule polymerization
          negative regulation of protein ubiquitination
          positive regulation of endodeoxyribonuclease activity
          DNA damage response, detection of DNA damage
          regulation of apoptotic process
          positive regulation of JUN kinase activity
          negative regulation of DNA repair
          positive regulation of DNA repair
          spindle assembly
          cell division
          response to TNF agonist
          cellular response to hydrogen peroxide
          positive regulation of NIK/NF-kappaB signaling
          positive regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          positive regulation of DNA N-glycosylase activity
          positive regulation of base-excision repair
          positive regulation of apoptotic signaling pathway
          positive regulation of cysteine-type endopeptidase activity involved in execution phase of apoptosis
          DNA binding
          damaged DNA binding
          RNA binding
          mRNA binding
          structural constituent of ribosome
          DNA-(apurinic or apyrimidinic site) lyase activity
          endodeoxyribonuclease activity
          protein binding
          microtubule binding
          transcription factor binding
          tubulin binding
          DNA N-glycosylase activity
          enzyme binding
          kinase binding
          protein kinase binding
          Hsp70 protein binding
          SUMO binding
          oxidized purine DNA binding
          oxidized pyrimidine DNA binding
          ubiquitin-like protein conjugating enzyme binding
          poly(A) RNA binding
          protein kinase A binding
          iron-sulfur cluster binding
          Hsp90 protein binding
          small ribosomal subunit rRNA binding
          supercoiled DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0