Search
Search

IGSF8| Gene Symbol: IGSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206665.2 | NP_001193594.1 | 7 | 1909 | ||
| NM_001320247.1 | NP_001307176.1 | 6 | 2051 | |||
| NM_052868.5 | NP_443100.1 | 6 | 2051 | |||
| XM_006711631.3 | XP_006711694.1 | 6 | 1800 | |||
| XM_011510159.2 | XP_011508461.1 | 6 | 2431 | |||
| XM_017002835.1 | XP_016858324.1 | 7 | 1915 | |||
| XM_017002836.1 | XP_016858325.1 | 7 | 2106 | |||
| XM_017002837.1 | XP_016858326.1 | 7 | 1915 | |||
| XM_017002838.1 | XP_016858327.1 | 6 | 4051 | |||
| XM_017002839.1 | XP_016858328.1 | 7 | 3963 | |||
| XM_017002840.1 | XP_016858329.1 | 7 | 2077 | |||
| GenBank mRNA | AF311906.1 | - | 7 | 1619 | ||
| AF407274.1 | - | 6 | 1991 | |||
| AK055843.1 | - | 6 | 1787 | |||
| AK090396.1 | - | 1 | 5186 | |||
| AK308115.1 | - | 7 | 1910 | |||
| AY044845.1 | - | 6 | 1906 | |||
| AY157579.1 | - | 6 | 1860 | |||
| BC004108.2 | - | 5 | 1448 | |||
| BC011896.2 | - | 3 | 929 | |||
| BC023649.2 | - | 4 | 1116 | |||
| BC053881.1 | - | 6 | 1997 | 5752209 | ||
| BU172537.1 | - | 2 | 156 | |||