Search
Search

FCRLB| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002901.3 | NP_001002901.1 | 7 | 941 | ||
| NM_001288831.1 | NP_001275760.1 | 5 | 623 | |||
| NM_001288832.1 | NP_001275761.1 | 4 | 602 | |||
| NM_001320241.1 | NP_001307170.1 | 6 | 755 | |||
| GenBank mRNA | AK055614.1 | - | 1 | 935 | ||
| AK290733.1 | - | 6 | 755 | |||
| AY670683.1 | - | 5 | 722 | |||
| AY670686.1 | - | 5 | 618 | |||
| AY670687.1 | - | 4 | 597 | |||
| AY786314.1 | - | 5 | 707 | |||
| BC038564.2 | - | 2 | 307 | |||
| BC067080.1 | - | 1 | 924 | |||
| DQ021957.1 | - | 7 | 941 | |||