Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CHD3 SIRNA Assays ›
Gene Symbol:
CHD3
Gene Name:
chromodomain helicase DNA binding protein 3
Gene Aliases:
Mi-2a, Mi2-ALPHA, ZFH
Chromosome Location:
Chr.17: 7884764 - 7912757 on Build GRCh38
UniGene ID:
Hs.25601
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
216241
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CHD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005271.2
NP_001005271.2
25, 26
4255
NM_001005273.2
NP_001005273.1
25, 26
4226
NM_005852.3
NP_005843.2
25, 26
4226
XM_005256427.4
XP_005256484.1
25, 26
4296
XM_005256428.4
XP_005256485.1
25, 26
4296
XM_005256429.4
XP_005256486.1
25, 26
4296
XM_006721423.3
XP_006721486.1
25, 26
4296
XM_006721424.3
XP_006721487.1
25, 26
4271
XM_006721428.3
XP_006721491.1
25, 26
4296
XM_017024061.1
XP_016879550.1
25, 26
4278
XM_017024062.1
XP_016879551.1
25, 26
4253
XM_017024063.1
XP_016879552.1
25, 26
4211
XM_017024064.1
XP_016879553.1
25, 26
4271
XM_017024065.1
XP_016879554.1
24, 25
4092
XM_017024066.1
XP_016879555.1
26, 27
4031
XM_017024067.1
XP_016879556.1
26, 27
4214
XM_017024068.1
XP_016879557.1
25, 26
4187
XM_017024069.1
XP_016879558.1
25, 26
4078
XM_017024070.1
XP_016879559.1
25, 26
4253
GenBank mRNA
AK125928.1
-
2, 3
639
AK296937.1
-
7, 8
1020
AK308362.1
-
3, 4
386
BC110648.1
-
1, 2
59
U91543.1
-
25, 26
4226
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
chromatin assembly or disassembly
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
spindle organization
histone deacetylation
DNA duplex unwinding
centrosome organization
regulation of signal transduction by p53 class mediator
DNA binding
ATP-dependent DNA helicase activity
helicase activity
histone deacetylase activity
protein binding
ATP binding
zinc ion binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0