Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPIP5K1 SIRNA Assays ›
          Gene Symbol:
          PPIP5K1
          Gene Name:
          diphosphoinositol pentakisphosphate kinase 1
          Gene Aliases:
          HISPPD2A, IP6K, IPS1, VIP1, hsVIP1
          Chromosome Location:
          Chr.15: 43533462 - 43590272 on Build GRCh38
          UniGene ID:
          Hs.156814
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 21662

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPIP5K1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130858.2 NP_001124330.1 3, 4 350
          NM_001130859.2 NP_001124331.1 2, 3 345
          NM_001190214.1 NP_001177143.1 3, 4 350
          NM_014659.5 NP_055474.3 2, 3 268
          XM_005254803.1 XP_005254860.1 3, 4 350
          XM_005254804.1 XP_005254861.1 3, 4 350
          XM_006720786.2 XP_006720849.1 3, 4 350
          XM_011522249.1 XP_011520551.1 3, 4 350
          XM_011522250.1 XP_011520552.1 3, 4 453
          XM_011522252.1 XP_011520554.1 2, 3 324
          XM_011522254.1 XP_011520556.1 2, 3 220
          XM_011522259.1 XP_011520561.1 3, 4 350
          XM_017022735.1 XP_016878224.1 2, 3 667
          XM_017022736.1 XP_016878225.1 2, 3 635
          XM_017022737.1 XP_016878226.1 2, 3 316
          XM_017022738.1 XP_016878227.1 2, 3 264
          XM_017022739.1 XP_016878228.1 2, 3 264
          XM_017022740.1 XP_016878229.1 3, 4 453
          XM_017022741.1 XP_016878230.1 2, 3 666
          XM_017022742.1 XP_016878231.1 3, 4 458
          XM_017022743.1 XP_016878232.1 2, 3 324
          XM_017022744.1 XP_016878233.1 2, 3 265
          XM_017022745.1 XP_016878234.1 3, 4 453
          XM_017022746.1 XP_016878235.1 2, 3 324
          XM_017022747.1 XP_016878236.1 2, 3 264
          XM_017022748.1 XP_016878237.1 2, 3 666
          XM_017022749.1 XP_016878238.1 3, 4 426
          XM_017022750.1 XP_016878239.1 2, 3 267
          XM_017022751.1 XP_016878240.1 2, 3 220
          XM_017022752.1 XP_016878241.1 3, 4 350
          XM_017022753.1 XP_016878242.1 2, 3 324
          XM_017022754.1 XP_016878243.1 3, 4 458
          XM_017022755.1 XP_016878244.1 2, 3 324
          XM_017022756.1 XP_016878245.1 2, 3 262
          XM_017022757.1 XP_016878246.1 2, 3 220
          XM_017022759.1 XP_016878248.1 2, 3 264
          XM_017022760.1 XP_016878249.1 2, 3 283
          GenBank mRNA AB002375.1 - 2, 3 272
          AF502586.1 - 2, 3 240
          AF502587.1 - 2, 3 272
          AF502588.1 - 2, 3 240
          AF502589.1 - 2, 3 272
          AF543190.1 - 2, 3 222
          AK307438.1 - 3, 4 314
          BC050263.1 - 1, 2 296 5521199
          BC057395.1 - 2, 3 296 6142663
          BX538022.1 - 2, 3 296
          BX647814.1 - 3, 4 305
          DB065605.1 - 3, 4 352
          DB228266.1 - 2, 3 324

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          nucleotide kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          inositol metabolic process
          phosphorylation
          dephosphorylation
          inositol phosphate biosynthetic process
          inositol phosphate metabolic process
          inositol-1,3,4,5,6-pentakisphosphate kinase activity
          inositol hexakisphosphate kinase activity
          inositol heptakisphosphate kinase activity
          inositol hexakisphosphate 5-kinase activity
          acid phosphatase activity
          ATP binding
          diphosphoinositol-pentakisphosphate kinase activity
          inositol hexakisphosphate 1-kinase activity
          inositol hexakisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0