Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DEPDC5 SIRNA Assays ›
          Gene Symbol:
          DEPDC5
          Gene Name:
          DEP domain containing 5
          Gene Aliases:
          DEP.5, FFEVF
          Chromosome Location:
          Chr.22: 31753951 - 31907034 on Build GRCh38
          UniGene ID:
          Hs.435022
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 21665

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DEPDC5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007188.2 NP_001007189.1 4, 5 387
          NM_001136029.2 NP_001129501.1 3, 4 245
          NM_001242896.1 NP_001229825.1 4, 5 387
          NM_001242897.1 NP_001229826.1 4, 5 291
          NM_014662.4 NP_055477.1 3, 4 245
          NR_110988.1 4, 5 387
          XM_005261862.1 XP_005261919.1 4, 5 291
          XM_011530557.2 XP_011528859.1 4, 5 394
          XM_011530558.2 XP_011528860.1 4, 5 394
          XM_011530559.2 XP_011528861.1 4, 5 394
          XM_011530560.2 XP_011528862.1 4, 5 413
          XM_011530561.2 XP_011528863.1 4, 5 394
          XM_011530562.2 XP_011528864.1 4, 5 394
          XM_011530563.2 XP_011528865.1 4, 5 394
          XM_011530565.2 XP_011528867.1 4, 5 394
          XM_011530568.2 XP_011528870.1 4, 5 394
          XM_017029109.1 XP_016884598.1 4, 5 291
          XM_017029110.1 XP_016884599.1 4, 5 291
          XM_017029111.1 XP_016884600.1 4, 5 291
          XM_017029112.1 XP_016884601.1 4, 5 291
          XM_017029113.1 XP_016884602.1 4, 5 394
          XM_017029114.1 XP_016884603.1 4, 5 394
          XM_017029115.1 XP_016884604.1 4, 5 394
          GenBank mRNA AB014545.1 - 3, 4 244
          AJ698950.1 - 3, 4 240
          AJ698951.1 - 3, 4 240
          AJ704764.1 - 3, 4 240
          AK294987.1 - 4, 5 263
          AK300737.1 - 4, 5 356
          BC057797.1 - 4, 5 327
          BC136612.1 - 4, 5 301 9020602
          BC146766.1 - 3, 4 244 9007209
          BP354669.1 - 4, 5 388
          DA339804.1 - 4, 5 291

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          G-protein modulator protein-binding activity modulator GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of TOR signaling
          cellular response to amino acid starvation
          intracellular signal transduction
          positive regulation of GTPase activity
          negative regulation of TORC1 signaling
          GTPase activator activity
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9c8cf:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0