Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAPS2 SIRNA Assays ›
          Gene Symbol:
          CAPS2
          Gene Name:
          calcyphosine 2
          Gene Aliases:
          UG0636c06
          Chromosome Location:
          Chr.12: 75275979 - 75390941 on Build GRCh38
          UniGene ID:
          Hs.733835
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 217057

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAPS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286547.1 NP_001273476.1 7, 8 761
          NM_001286548.1 NP_001273477.1 8, 9 1038
          NM_001286549.1 NP_001273478.1 4, 5 478
          NM_032606.3 NP_115995.2 8, 9 910
          XM_005269192.1 XP_005269249.1 6, 7 589
          XM_005269194.3 XP_005269251.1 2, 3 216
          XM_006719648.2 XP_006719711.2 8, 9 910
          XM_006719649.2 XP_006719712.2 8, 9 910
          XM_011538878.1 XP_011537180.1 8, 9 875
          XM_011538886.2 XP_011537188.1 7, 8 844
          XM_011538887.2 XP_011537189.1 7, 8 844
          XM_011538888.1 XP_011537190.1 4, 5 429
          XM_011538889.2 XP_011537191.1 8, 9 781
          XM_011538890.1 XP_011537192.1 8, 9 910
          XM_011538892.1 XP_011537194.1 5, 6 475
          XM_017020034.1 XP_016875523.1 9, 10 833
          XM_017020035.1 XP_016875524.1 4, 5 429
          XM_017020036.1 XP_016875525.1 9, 10 1000
          XM_017020037.1 XP_016875526.1 6, 7 629
          XM_017020038.1 XP_016875527.1 4, 5 497
          XM_017020039.1 XP_016875528.1 3, 4 293
          XM_017020041.1 XP_016875530.1 9, 10 1000
          XM_017020042.1 XP_016875531.1 9, 10 1028
          XM_017020043.1 XP_016875532.1 5, 6 845
          XM_017020044.1 XP_016875533.1 4, 5 439
          XM_017020045.1 XP_016875534.1 5, 6 881
          XM_017020046.1 XP_016875535.1 4, 5 438
          GenBank mRNA AF251056.1 - 8, 9 1038
          AF351611.1 - 7, 8 619
          AK091839.1 - 8, 9 910
          AK092274.1 - 2, 3 126
          BC033860.1 - 7, 8 730
          BC056672.1 - 3, 4 286 6616433
          BC131724.1 - 4, 5 477

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Process(es)

          calcium ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0