Search
Search

KANK2| Gene Symbol: KANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136191.2 | NP_001129663.1 | 10 | 2520 | ||
| NM_015493.6 | NP_056308.3 | 8 | 2262 | |||
| GenBank mRNA | AB040951.2 | - | 9 | 2472 | ||
| AB284125.1 | - | 11 | 2703 | |||
| AK002094.1 | - | 7 | 952 | |||
| AK023332.1 | - | 7 | 1004 | |||
| AY639929.1 | - | 8 | 2183 | |||
| BC082762.1 | - | 10 | 2474 | 6497026 | ||
| BC098105.1 | - | 9 | 2272 | 40012030 | ||
| BC098286.1 | - | 9 | 2272 | 40012033 | ||
| BC098312.1 | - | 9 | 2272 | 40012032 | ||