Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS4X SIRNA Assays ›
          Gene Symbol:
          Rps4x LOC100362640
          Gene Name:
          ribosomal protein S4, X-linked
          ribosomal protein S4, X-linked X-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.4: 35729619 - 35730512 on Build Rnor_6.0
          Chromosome Location:
          Chr.X: 72074108 - 72078576 on Build Rnor_6.0
          UniGene ID:
          Rn.40171
          Rn.40171
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 217852

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps4x
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007600.1 NP_001007601.1 1 507
          GenBank mRNA BC086560.1 - 1 507 7303578
          FQ220060.1 - 1 529
          FQ226324.1 - 1 530
          FQ230280.1 - 1 530
          X14210.1 - 1 528
          Gene Symbol: LOC100362640
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_002730258.3 XP_002730304.1 5 595
          GenBank mRNA FQ209633.1 - 1 119
          FQ210628.1 - 1 119
          FQ215274.1 - 1 119
          FQ217297.1 - 1 119
          FQ217469.1 - 1 119
          FQ218404.1 - 1 119
          FQ219331.1 - 1 119
          FQ219418.1 - 1 119
          FQ219965.1 - 1 119
          FQ219969.1 - 1 119
          FQ220033.1 - 1 119
          FQ220065.1 - 1 119
          FQ220251.1 - 1 119
          FQ220262.1 - 1 119
          FQ220354.1 - 1 119
          FQ220409.1 - 1 119
          FQ220511.1 - 1 119
          FQ220516.1 - 1 119
          FQ220573.1 - 1 119
          FQ220754.1 - 1 119
          FQ220755.1 - 1 119
          FQ220791.1 - 1 119
          FQ220859.1 - 1 119
          FQ220870.1 - 1 119
          FQ221047.1 - 1 119
          FQ221391.1 - 1 119
          FQ221464.1 - 1 119
          FQ222067.1 - 1 119
          FQ222104.1 - 1 119
          FQ222211.1 - 1 119
          FQ222355.1 - 1 119
          FQ222933.1 - 1 119
          FQ223416.1 - 1 119
          FQ224981.1 - 1 119
          FQ225568.1 - 1 119
          FQ226419.1 - 1 119
          FQ226822.1 - 1 119
          FQ227084.1 - 1 119
          FQ227791.1 - 1 119
          FQ228063.1 - 1 119
          FQ228315.1 - 1 119
          FQ229145.1 - 1 119
          FQ229151.1 - 1 119
          FQ229207.1 - 1 119
          FQ229260.1 - 1 119
          FQ229529.1 - 1 119
          FQ229631.1 - 1 119
          FQ229639.1 - 1 115
          FQ229644.1 - 1 119
          FQ229784.1 - 1 119
          FQ229794.1 - 1 119
          FQ229917.1 - 1 119
          FQ230007.1 - 1 119
          FQ230010.1 - 1 119
          FQ230025.1 - 1 119
          FQ230069.1 - 1 119
          FQ230153.1 - 1 119
          FQ230175.1 - 1 119
          FQ231759.1 - 1 119
          FQ232729.1 - 1 119
          FQ232779.1 - 1 119
          FQ232793.1 - 1 119
          FQ233106.1 - 1 119
          FQ234125.1 - 1 119

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          multicellular organism development
          biological_process
          positive regulation of cell proliferation
          positive regulation of translation
          molecular_function
          structural constituent of ribosome
          rRNA binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0