Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPLP1 SIRNA Assays ›
          Gene Symbol:
          Rplp1
          Gene Name:
          ribosomal protein, large, P1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 66862143 - 66863476 on Build Rnor_6.0
          UniGene ID:
          Rn.973
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 217864

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rplp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007604.2 NP_001007605.1 2 240
          GenBank mRNA BC058151.1 - 2 243 6888011
          FQ209966.1 - 2 242
          FQ210237.1 - 2 239
          FQ210632.1 - 2 241
          FQ210864.1 - 2 239
          FQ210988.1 - 2 242
          FQ211097.1 - 2 242
          FQ211327.1 - 2 240
          FQ211648.1 - 2 239
          FQ212351.1 - 2 239
          FQ214973.1 - 2 242
          FQ215065.1 - 2 239
          FQ216905.1 - 2 243
          FQ216921.1 - 2 242
          FQ216959.1 - 2 238
          FQ216979.1 - 2 242
          FQ217216.1 - 2 242
          FQ217313.1 - 2 240
          FQ220167.1 - 2 240
          FQ220212.1 - 2 241
          FQ220998.1 - 2 239
          FQ221027.1 - 2 240
          FQ221120.1 - 2 240
          FQ221137.1 - 2 239
          FQ221201.1 - 2 242
          FQ221359.1 - 2 240
          FQ221455.1 - 2 244
          FQ221534.1 - 2 237
          FQ221567.1 - 2 242
          FQ221595.1 - 2 240
          FQ221620.1 - 2 239
          FQ221698.1 - 2 246
          FQ221717.1 - 2 239
          FQ221756.1 - 2 239
          FQ221784.1 - 2 239
          FQ221894.1 - 2 239
          FQ221969.1 - 2 239
          FQ221980.1 - 2 239
          FQ222031.1 - 2 239
          FQ222178.1 - 2 239
          FQ222307.1 - 2 241
          FQ222468.1 - 2 241
          FQ222559.1 - 2 239
          FQ222608.1 - 2 242
          FQ222618.1 - 2 242
          FQ222784.1 - 2 239
          FQ222801.1 - 2 240
          FQ222805.1 - 2 239
          FQ222816.1 - 2 240
          FQ222879.1 - 2 235
          FQ222923.1 - 2 240
          FQ222965.1 - 2 239
          FQ222966.1 - 2 238
          FQ223013.1 - 2 240
          FQ223016.1 - 2 240
          FQ223066.1 - 2 241
          FQ223344.1 - 2 240
          FQ223388.1 - 2 239
          FQ223390.1 - 2 240
          FQ223470.1 - 2 242
          FQ223512.1 - 2 240
          FQ223537.1 - 2 240
          FQ223567.1 - 2 239
          FQ223591.1 - 2 239
          FQ223674.1 - 2 221
          FQ223769.1 - 2 240
          FQ224482.1 - 2 209
          FQ224625.1 - 2 242
          FQ224766.1 - 2 240
          FQ228323.1 - 2 239
          FQ228361.1 - 2 239
          FQ228390.1 - 2 240
          FQ228490.1 - 2 240
          FQ228523.1 - 2 239
          FQ228528.1 - 2 236
          FQ228539.1 - 2 239
          FQ228605.1 - 2 243
          FQ228650.1 - 2 239
          FQ228792.1 - 2 242
          FQ228990.1 - 2 246
          FQ229009.1 - 2 239
          FQ229050.1 - 2 239
          FQ229318.1 - 2 240
          FQ230089.1 - 2 240
          X15097.1 - 2 210

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_018853, AK010656, U29402, BC058685, BC092088, and others.

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          structural constituent of ribosome

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sztk8:80/100.66.77.162:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0