Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP6R2 SIRNA Assays ›
          Gene Symbol:
          PPP6R2
          Gene Name:
          protein phosphatase 6 regulatory subunit 2
          Gene Aliases:
          KIAA0685, PP6R2, SAP190, SAPS2
          Chromosome Location:
          Chr.22: 50343317 - 50445090 on Build GRCh38
          UniGene ID:
          Hs.449098
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 21864

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPP6R2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242898.1 NP_001229827.1 3 494
          NM_001242899.1 NP_001229828.1 3 494
          NM_001242900.1 NP_001229829.1 3 494
          NM_014678.4 NP_055493.2 3 494
          XM_006724431.2 XP_006724494.1 4 605
          XM_006724434.1 XP_006724497.1 5 754
          XM_006724435.2 XP_006724498.1 3 479
          XM_011530720.2 XP_011529022.1 2 179
          XM_011530721.2 XP_011529023.1 2 179
          XM_011530722.2 XP_011529024.1 2 179
          XM_011530723.2 XP_011529025.1 2 179
          XM_011530724.2 XP_011529026.1 2 179
          XM_011530726.2 XP_011529028.1 2 179
          XM_011530727.2 XP_011529029.1 2 179
          XM_011530728.2 XP_011529030.1 2 179
          XM_011530729.2 XP_011529031.1 2 179
          XM_011530730.2 XP_011529032.1 2 179
          XM_011530731.1 XP_011529033.1 2 179
          XM_011530732.2 XP_011529034.1 5 655
          XM_011530734.2 XP_011529036.1 4 529
          XM_011530735.2 XP_011529037.1 3 479
          XM_011530736.2 XP_011529038.1 4 605
          XM_011530737.2 XP_011529039.1 5 754
          XM_011530738.2 XP_011529040.1 4 557
          XM_011530739.2 XP_011529041.1 2 219
          XM_011530740.2 XP_011529042.1 4 605
          XM_017029116.1 XP_016884605.1 2 179
          XM_017029117.1 XP_016884606.1 6 804
          XM_017029118.1 XP_016884607.1 6 756
          XM_017029119.1 XP_016884608.1 5 678
          XM_017029120.1 XP_016884609.1 5 655
          XM_017029121.1 XP_016884610.1 3 479
          XM_017029122.1 XP_016884611.1 4 557
          XM_017029123.1 XP_016884612.1 3 479
          XM_017029124.1 XP_016884613.1 4 605
          XM_017029125.1 XP_016884614.1 3 479
          XM_017029126.1 XP_016884615.1 3 479
          XM_017029127.1 XP_016884616.1 4 557
          XM_017029128.1 XP_016884617.1 4 605
          XM_017029129.1 XP_016884618.1 3 479
          XM_017029130.1 XP_016884619.1 3 479
          XM_017029131.1 XP_016884620.1 4 605
          XM_017029132.1 XP_016884621.1 4 557
          XM_017029133.1 XP_016884622.1 3 479
          XM_017029134.1 XP_016884623.1 3 479
          XM_017029135.1 XP_016884624.1 4 529
          XM_017029136.1 XP_016884625.1 3 479
          XM_017029137.1 XP_016884626.1 3 479
          XM_017029138.1 XP_016884627.1 3 479
          GenBank mRNA AB014585.1 - 3 466
          AK302472.1 - 3 405
          BC000976.4 - 4 483 2900437
          BC006568.2 - 3 480 3532211
          BC032664.1 - 3 491 5553142
          BC041698.1 - 3 461 5732101
          BC052995.2 - 3 481 6162781

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0