Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDE2A SIRNA Assays ›
          Gene Symbol:
          Pde2a
          Gene Name:
          phosphodiesterase 2A, cGMP-stimulated
          Gene Aliases:
          CGS-PDE, cGSPDE
          Chromosome Location:
          Chr.7: 101421691 - 101512829 on Build GRCm38
          UniGene ID:
          Mm.247564
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 218802

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pde2a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008548.4 NP_001008548.1 31 3496
          NM_001143848.2 NP_001137320.1 31 3508
          NM_001143849.2 NP_001137321.1 30 3341
          NM_001243757.1 NP_001230686.1 31 3403
          NM_001243758.1 NP_001230687.1 31 3400
          GenBank mRNA AK158560.1 - 31 3401
          AK158888.1 - 22 2384
          AK159012.1 - 31 3290
          AK170395.1 - 31 3280
          AK170573.1 - 30 3280
          AK172397.1 - 30 3282
          BC006845.1 - 11 1554
          BC029810.1 - 15 1998
          BC057029.1 - 32 3402
          BC086800.1 - 31 3478 30543765

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phosphodiesterase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cAMP catabolic process
          protein targeting to mitochondrion
          signal transduction
          metabolic process
          positive regulation of gene expression
          cAMP-mediated signaling
          cGMP-mediated signaling
          negative regulation of cAMP biosynthetic process
          regulation of cGMP metabolic process
          negative regulation of protein import into nucleus, translocation
          cellular response to drug
          cellular response to macrophage colony-stimulating factor stimulus
          negative regulation of vascular permeability
          positive regulation of vascular permeability
          cGMP catabolic process
          positive regulation of inflammatory response
          establishment of endothelial barrier
          cellular response to mechanical stimulus
          cellular response to cGMP
          cellular response to granulocyte macrophage colony-stimulating factor stimulus
          nucleotide binding
          cyclic-nucleotide phosphodiesterase activity
          3',5'-cyclic-nucleotide phosphodiesterase activity
          cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
          phosphoric diester hydrolase activity
          drug binding
          hydrolase activity
          cAMP binding
          cGMP binding
          TPR domain binding
          GAF domain binding
          protein homodimerization activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0