Search
Search

Chfr| Gene Symbol: Chfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009258.1 | NP_001009258.1 | 17, 18 | 2003 | ||
| XM_006249535.2 | XP_006249597.1 | 17, 18 | 2085 | |||
| XM_006249536.2 | XP_006249598.1 | 16, 17 | 1965 | |||
| GenBank mRNA | BC087162.1 | - | 17, 18 | 2003 | 7135077 | |