Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NFASC SIRNA Assays ›
          Gene Symbol:
          NFASC
          Gene Name:
          neurofascin
          Gene Aliases:
          NF, NRCAML
          Chromosome Location:
          Chr.1: 204828652 - 205022822 on Build GRCh38
          UniGene ID:
          Hs.13349
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 220924

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NFASC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005388.2 NP_001005388.2 1, 2 137
          NM_001005389.1 NP_001005389.2 1, 2 137
          NM_001160332.1 NP_001153804.1 1, 2 137
          NM_001160333.1 NP_001153805.1 1, 2 137
          NM_015090.3 NP_055905.2 1, 2 137
          XM_005244989.3 XP_005245046.2 1, 2 217
          XM_005244991.3 XP_005245048.2 1, 2 217
          XM_005244992.4 XP_005245049.2 1, 2 217
          XM_005244993.3 XP_005245050.2 1, 2 217
          XM_005244997.3 XP_005245054.2 1, 2 217
          XM_011509312.1 XP_011507614.1 1, 2 217
          XM_011509314.1 XP_011507616.1 1, 2 217
          XM_011509316.1 XP_011507618.1 1, 2 96
          XM_011509318.2 XP_011507620.1 1, 2 139
          XM_011509319.1 XP_011507621.1 1, 2 96
          XM_011509320.2 XP_011507622.1 1, 2 139
          XM_017000733.1 XP_016856222.1 1, 2 217
          XM_017000734.1 XP_016856223.1 1, 2 217
          XM_017000738.1 XP_016856227.1 1, 2 139
          XM_017000739.1 XP_016856228.1 1, 2 139
          XM_017000740.1 XP_016856229.1 1, 2 139
          XM_017000743.1 XP_016856232.1 1, 2 139
          GenBank mRNA AK090639.1 - 1, 2 96
          AK295291.1 - 1, 2 116
          AK299262.1 - 1, 2 116
          AK308686.1 - 1, 2 217
          BC040674.1 - 1, 2 142
          BG702675.1 - 1, 2 142

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein localization to paranode region of axon
          axon guidance
          peripheral nervous system development
          transmission of nerve impulse
          paranodal junction assembly
          heterotypic cell-cell adhesion
          myelination
          clustering of voltage-gated sodium channels
          synapse organization
          protein localization to juxtaparanode region of axon
          protein targeting to plasma membrane
          protein binding
          protein domain specific binding
          protein binding involved in heterotypic cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gh6bl:80/100.66.76.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0