Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CST3 SIRNA Assays ›
          Gene Symbol:
          Cst3
          Gene Name:
          cystatin C
          Gene Aliases:
          CYSC
          Chromosome Location:
          Chr.3: 143219671 - 143223544 on Build Rnor_6.0
          UniGene ID:
          Rn.106351
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 221324

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cst3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012837.1 NP_036969.1 3 519
          GenBank mRNA BC087591.1 - 3 459 7307694
          CB314369.1 - 3 291
          CB608544.1 - 3 502
          FQ210306.1 - 3 472
          FQ211221.1 - 3 472
          FQ211526.1 - 3 472
          FQ211840.1 - 3 472
          FQ211941.1 - 3 472
          FQ212057.1 - 3 501
          FQ212112.1 - 3 472
          FQ212277.1 - 3 472
          FQ212367.1 - 3 472
          FQ212374.1 - 3 472
          FQ214337.1 - 3 472
          FQ218053.1 - 3 503
          FQ221232.1 - 3 472
          FQ221782.1 - 3 472
          FQ222268.1 - 3 472
          FQ222817.1 - 2 153
          FQ222820.1 - 3 472
          FQ223224.1 - 3 472
          FQ224103.1 - 3 472
          FQ228332.1 - 3 469
          FQ228399.1 - 3 472
          FQ228646.1 - 3 472
          FQ234324.1 - 3 503
          X16957.1 - 3 413

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          eye development
          response to hypoxia
          cell activation
          apoptotic process
          defense response
          response to oxidative stress
          brain development
          salivary gland development
          embryo implantation
          positive regulation of cell proliferation
          male gonad development
          response to toxic substance
          response to carbohydrate
          response to inorganic substance
          negative regulation of peptidase activity
          negative regulation of collagen catabolic process
          negative regulation of extracellular matrix disassembly
          negative regulation of endopeptidase activity
          response to organic cyclic compound
          response to nutrient levels
          response to estradiol
          regulation of tissue remodeling
          cellular response to oxidative stress
          response to drug
          circadian sleep/wake cycle, REM sleep
          regulation of programmed cell death
          extracellular fibril organization
          positive regulation of DNA replication
          negative regulation of proteolysis
          response to axon injury
          Sertoli cell development
          negative regulation of elastin catabolic process
          negative regulation of blood vessel remodeling
          negative regulation of cell death
          cellular response to hydrogen peroxide
          beta-amyloid binding
          protease binding
          endopeptidase inhibitor activity
          cysteine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0