Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNAP91 SIRNA Assays ›
Gene Symbol:
SNAP91
Gene Name:
synaptosome associated protein 91
Gene Aliases:
AP180, CALM
Chromosome Location:
Chr.6: 83552885 - 83709408 on Build GRCh38
UniGene ID:
Hs.368046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22170
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNAP91
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242792.1
NP_001229721.1
3
553
NM_001242793.1
NP_001229722.1
3
352
NM_001242794.1
NP_001229723.1
3
352
NM_001256717.1
NP_001243646.1
6
788
NM_014841.2
NP_055656.1
3
556
XM_005248770.4
XP_005248827.1
3
395
XM_006715615.1
XP_006715678.1
3
347
XM_011536265.1
XP_011534567.1
3
354
XM_011536266.1
XP_011534568.1
3
466
XM_011536269.1
XP_011534571.1
3
347
XM_011536271.1
XP_011534573.1
3
347
XM_011536273.2
XP_011534575.1
3
347
XM_011536275.2
XP_011534577.1
3
347
XM_011536276.2
XP_011534578.1
3
347
XM_011536277.1
XP_011534579.1
5
635
XM_017011554.1
XP_016867043.1
3
511
XM_017011555.1
XP_016867044.1
3
395
XM_017011556.1
XP_016867045.1
3
327
XM_017011557.1
XP_016867046.1
3
524
XM_017011558.1
XP_016867047.1
3
354
XM_017011559.1
XP_016867048.1
3
453
XM_017011560.1
XP_016867049.1
3
395
XM_017011561.1
XP_016867050.1
3
466
XM_017011562.1
XP_016867051.1
3
524
XM_017011563.1
XP_016867052.1
3
524
XM_017011564.1
XP_016867053.1
3
395
XM_017011565.1
XP_016867054.1
3
453
XM_017011566.1
XP_016867055.1
3
524
XM_017011567.1
XP_016867056.1
3
327
XM_017011568.1
XP_016867057.1
3
524
XM_017011569.1
XP_016867058.1
3
524
XM_017011570.1
XP_016867059.1
3
524
XM_017011571.1
XP_016867060.1
3
395
XM_017011572.1
XP_016867061.1
3
354
XM_017011573.1
XP_016867062.1
3
453
XM_017011574.1
XP_016867063.1
3
524
XM_017011575.1
XP_016867064.1
3
524
XM_017011576.1
XP_016867065.1
3
327
XM_017011577.1
XP_016867066.1
3
453
XM_017011578.1
XP_016867067.1
3
466
XM_017011579.1
XP_016867068.1
3
524
XM_017011580.1
XP_016867069.1
3
524
XM_017011581.1
XP_016867070.1
3
354
XM_017011582.1
XP_016867071.1
3
383
XM_017011583.1
XP_016867072.1
3
511
XM_017011584.1
XP_016867073.1
3
354
XM_017011585.1
XP_016867074.1
3
395
XM_017011586.1
XP_016867075.1
3
347
XM_017011587.1
XP_016867076.1
3
453
XM_017011588.1
XP_016867077.1
3
466
XM_017011589.1
XP_016867078.1
3
524
XM_017011590.1
XP_016867079.1
3
395
GenBank mRNA
AB014556.1
-
3
482
AK289582.1
-
3
524
BC060818.1
-
3
336
4827211
CR749348.1
-
6
755
DA307867.1
-
3
407
DA316497.1
-
3
347
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
protein transport
clathrin coat assembly
regulation of clathrin-mediated endocytosis
protein binding
1-phosphatidylinositol binding
protein kinase binding
clathrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0