Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMG7 SIRNA Assays ›
          Gene Symbol:
          SMG7
          Gene Name:
          SMG7, nonsense mediated mRNA decay factor
          Gene Aliases:
          C1orf16, EST1C, SGA56M
          Chromosome Location:
          Chr.1: 183471962 - 183554193 on Build GRCh38
          UniGene ID:
          Hs.591463
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 22264

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMG7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001174061.1 NP_001167532.1 18, 19 3199
          NM_173156.2 NP_775179.1 18, 19 3081
          NM_201568.2 NP_963862.1 18, 19 2943
          NM_201569.2 NP_963863.2 19, 20 3093
          XM_005245648.3 XP_005245705.1 21, 22 3916
          XM_005245649.3 XP_005245706.1 20, 21 3779
          XM_005245652.3 XP_005245709.1 19, 20 3100
          XM_005245653.4 XP_005245710.1 20, 21 3202
          XM_006711676.3 XP_006711739.1 20, 21 3641
          XM_006711678.3 XP_006711741.1 19, 20 2962
          XM_011510205.2 XP_011508507.1 22, 23 4094
          XM_011510206.2 XP_011508508.1 22, 23 3956
          XM_011510207.2 XP_011508509.1 20, 21 3818
          XM_011510208.2 XP_011508510.1 21, 22 3944
          XM_011510209.2 XP_011508511.1 21, 22 3806
          XM_011510210.2 XP_011508512.1 18, 19 2932
          XM_017002966.1 XP_016858455.1 21, 22 3233
          XM_017002967.1 XP_016858456.1 20, 21 3091
          XM_017002968.1 XP_016858457.1 17, 18 2786
          XM_017002969.1 XP_016858458.1 19, 20 3293
          XM_017002970.1 XP_016858459.1 19, 20 3217
          XM_017002971.1 XP_016858460.1 19, 20 3155
          GenBank mRNA AB085674.1 - 18, 19 2894
          AK299178.1 - 18, 19 3070
          AL833675.1 - 6, 7 1326
          AY386363.1 - 11, 12 2050
          BC036381.1 - 19, 20 2961 4829267
          BC052565.1 - 11, 12 1938
          D87437.2 - 18, 19 2787

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          mRNA export from nucleus
          regulation of dephosphorylation
          protein binding
          telomeric DNA binding
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0