Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD6 SIRNA Assays ›
          Gene Symbol:
          ANKRD6
          Gene Name:
          ankyrin repeat domain 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 89433178 - 89633834 on Build GRCh38
          UniGene ID:
          Hs.702213
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 22343

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242809.1 NP_001229738.1 4, 5 667
          NM_001242811.1 NP_001229740.1 4, 5 1050
          NM_001242813.1 NP_001229742.1 4, 5 667
          NM_014942.4 NP_055757.3 4, 5 667
          XM_005248679.3 XP_005248736.1 3, 4 535
          XM_005248680.3 XP_005248737.1 5, 6 2839
          XM_005248681.2 XP_005248738.1 4, 5 768
          XM_005248682.3 XP_005248739.1 3, 4 535
          XM_005248683.3 XP_005248740.1 3, 4 535
          XM_005248684.3 XP_005248741.1 3, 4 535
          XM_011535596.2 XP_011533898.1 3, 4 535
          XM_011535598.2 XP_011533900.1 3, 4 537
          XM_017010493.1 XP_016865982.1 3, 4 535
          XM_017010494.1 XP_016865983.1 3, 4 535
          XM_017010495.1 XP_016865984.1 4, 5 636
          XM_017010496.1 XP_016865985.1 3, 4 701
          XM_017010497.1 XP_016865986.1 3, 4 469
          XM_017010498.1 XP_016865987.1 4, 5 636
          XM_017010499.1 XP_016865988.1 3, 4 535
          XM_017010500.1 XP_016865989.1 3, 4 535
          XM_017010501.1 XP_016865990.1 4, 5 636
          XM_017010502.1 XP_016865991.1 3, 4 2218
          XM_017010504.1 XP_016865993.1 3, 4 469
          XM_017010506.1 XP_016865995.1 3, 4 469
          XM_017010508.1 XP_016865997.1 3, 4 2213
          XM_017010509.1 XP_016865998.1 3, 4 469
          XM_017010510.1 XP_016865999.1 3, 4 524
          XM_017010511.1 XP_016866000.1 3, 4 2227
          XM_017010512.1 XP_016866001.1 4, 5 636
          XM_017010513.1 XP_016866002.1 4, 5 768
          XM_017010514.1 XP_016866003.1 3, 4 537
          XM_017010515.1 XP_016866004.1 3, 4 469
          GenBank mRNA AB023174.1 - 4, 5 494
          AK295731.1 - 4, 5 967
          AK307792.1 - 4, 5 636
          AL117504.1 - 1, 2 68
          BC042173.1 - 4, 5 632 5454509

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein-binding activity modulator kinase inhibitor kinase modulator non-receptor serine/threonine protein kinase ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s)

          positive regulation of JNK cascade
          negative regulation of canonical Wnt signaling pathway
          positive regulation of Wnt signaling pathway, planar cell polarity pathway

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0