Search
Search

| Gene Symbol: CCDC103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258395.1 | NP_001245324.1 | 4 | 711 | ||
| NM_001258396.1 | NP_001245325.1 | 4 | 725 | |||
| NM_001258397.1 | NP_001245326.1 | 4 | 788 | |||
| NM_001258398.1 | NP_001245327.1 | 4 | 726 | |||
| NM_001258399.1 | NP_001245328.1 | 4 | 745 | |||
| NM_213607.2 | NP_998772.1 | 4 | 741 | |||
| GenBank mRNA | AK289760.1 | - | 4 | 671 | ||
| BC041060.1 | - | 4 | 667 | 5455678 | ||
| Gene Symbol: FAM187A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023156.1 | - | 4 | 679 | ||