Search
Search

Cox14| Gene Symbol: Cox14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183256.3 | NP_899079.1 | 2 | 288 | ||
| GenBank mRNA | AK002816.1 | - | 2 | 151 | ||
| AK004378.1 | - | 2 | 107 | |||
| AK009401.1 | - | 2 | 260 | |||
| BC059905.1 | - | 2 | 240 | 6823331 | ||
| BC092142.1 | - | 2 | 189 | 6814201 | ||
| BP774190.1 | - | 2 | 315 | |||