Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PUM2 SIRNA Assays ›
Gene Symbol:
PUM2
Gene Name:
pumilio RNA binding family member 2
Gene Aliases:
PUMH2, PUML2
Chromosome Location:
Chr.2: 20248692 - 20352423 on Build GRCh38
UniGene ID:
Hs.467824
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22772
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PUM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282752.1
NP_001269681.1
3, 4
510
NM_001282790.1
NP_001269719.1
3, 4
373
NM_001282791.1
NP_001269720.1
4, 5
498
NM_015317.2
NP_056132.1
4, 5
619
XM_005262607.1
XP_005262664.1
4, 5
593
XM_005262609.1
XP_005262666.1
4, 5
593
XM_005262610.1
XP_005262667.1
4, 5
593
XM_006711973.1
XP_006712036.1
4, 5
390
XM_006711974.2
XP_006712037.1
4, 5
430
XM_006711975.2
XP_006712038.1
5, 6
554
XM_011532719.1
XP_011531021.1
6, 7
789
XM_011532720.2
XP_011531022.1
6, 7
798
XM_011532721.2
XP_011531023.1
5, 6
590
XM_011532724.2
XP_011531026.1
4, 5
593
XM_011532725.2
XP_011531027.1
4, 5
593
XM_017003701.1
XP_016859190.1
4, 5
390
XM_017003702.1
XP_016859191.1
4, 5
430
XM_017003703.1
XP_016859192.1
5, 6
618
XM_017003704.1
XP_016859193.1
7, 8
936
XM_017003705.1
XP_016859194.1
7, 8
923
XM_017003706.1
XP_016859195.1
6, 7
715
XM_017003707.1
XP_016859196.1
4, 5
390
XM_017003708.1
XP_016859197.1
4, 5
623
XM_017003709.1
XP_016859198.1
5, 6
589
XM_017003710.1
XP_016859199.1
4, 5
439
XM_017003711.1
XP_016859200.1
4, 5
390
XM_017003712.1
XP_016859201.1
4, 5
458
XM_017003713.1
XP_016859202.1
5, 6
617
GenBank mRNA
AF272350.1
-
1, 2
20
AF315591.1
-
3, 4
370
AK093847.1
-
4, 5
471
AK304198.1
-
3, 4
280
BC112046.1
-
3, 4
373
8327514
BC112048.1
-
3, 4
373
8327516
BC143550.1
-
3, 4
373
9052061
BC143552.1
-
4, 5
498
D87078.3
-
3, 4
347
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
posttranscriptional regulation of gene expression
stress granule assembly
regulation of mRNA stability
regulation of chromosome segregation
regulation of gene silencing by miRNA
positive regulation of RIG-I signaling pathway
positive regulation of gene silencing by miRNA
RNA binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0